Skip to Content
Merck
All Photos(1)

Key Documents

143480

Sigma-Aldrich

3-Aminothiophenol

96%

Synonym(s):

3-Aminobenzenethiol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4SH
CAS Number:
Molecular Weight:
125.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

96%

form

liquid

refractive index

n20/D 1.656 (lit.)

density

1.179 g/mL at 25 °C (lit.)

SMILES string

Nc1cccc(S)c1

InChI

1S/C6H7NS/c7-5-2-1-3-6(8)4-5/h1-4,8H,7H2

InChI key

KFFUEVDMVNIOHA-UHFFFAOYSA-N

Application

3-Aminothiophenol was used as analyte during the fabrication of top-to-bottom joined system consisting of a silver nanowire and nanospheres[1]. It was used in formation of polyaniline-analog monolayer on gold surface by anchoring to the gold substrate through the thiol moiety[2].

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evaluation of coverage of self-assembled polyaniline-analogue monolayer by capacitance measurements.
Niu L, et al.
Journal of Electroanalytical Chemistry, 600(1), 95-102 (2007)
Seongyong Kim et al.
Physical chemistry chemical physics : PCCP, 15(12), 4146-4153 (2012-11-21)
A top-to-bottom joined system consisting of a silver nanowire and nanospheres was fabricated by embedding silver nanospheres on a glass or silicon substrate on which 3-aminothiophenol as an analyte molecule was adsorbed, and then placing silver nanowires on the substrate
Luca Quattrini et al.
Scientific reports, 9(1), 9943-9943 (2019-07-11)
Melanoma is the most serious form of skin cancer but its medication is still far from being safe and thoroughly effective. The search of novel therapeutic approaches represents therefore a health emergency to push through eagerly. In this study, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service