Skip to Content
Merck
All Photos(1)

Key Documents

141712

Sigma-Aldrich

trans-1,2-Cyclohexanediol

98%

Synonym(s):

1,2-trans -Cyclohexanediol, 1,2-trans -Dihydroxycyclohexane, trans -2-Hydroxycyclohexanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H10(OH)2
CAS Number:
Molecular Weight:
116.16
Beilstein:
3193810
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

form

solid

mp

101-104 °C (lit.)

SMILES string

O[C@@H]1CCCC[C@H]1O

InChI

1S/C6H12O2/c7-5-3-1-2-4-6(5)8/h5-8H,1-4H2/t5-,6-/m1/s1

InChI key

PFURGBBHAOXLIO-PHDIDXHHSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Lisa D Cervia et al.
Molecular therapy. Nucleic acids, 11, 263-271 (2018-06-03)
The nuclear envelope is a physiological barrier to electrogene transfer. To understand different mechanisms of the nuclear entry for electrotransfected plasmid DNA (pDNA), the current study investigated how manipulation of the mechanisms could affect electrotransfection efficiency (eTE), transgene expression level
J Mráz et al.
Scandinavian journal of work, environment & health, 25(3), 233-237 (1999-08-18)
This study explored the acute effect of ethanol (EtOH) on the urinary excretion of cyclohexanol (CH-ol), 1,2- and 1,4-cyclohexanediol (CH-diol), biomarkers of exposure to important solvents, and chemical intermediates cyclohexanone (CH-one), cyclohexane (CH) and cyclohexanol. Volunteers (5-8 in each group)
Yoshihito Shiota et al.
Inorganic chemistry, 50(13), 6200-6209 (2011-06-04)
The catalytic conversion of 1,2-cyclohexanediol to adipic anhydride by Ru(IV)O(tpa) (tpa ═ tris(2-pyridylmethyl)amine) is discussed using density functional theory calculations. The whole reaction is divided into three steps: (1) formation of α-hydroxy cyclohexanone by dehydrogenation of cyclohexanediol, (2) formation of
M Tanaka et al.
The Journal of organic chemistry, 66(8), 2667-2673 (2001-04-17)
Diastereoselective alkylation of ethyl 2-methyl- and/or 2-ethylacetoacetates using the (S,S)-cyclohexane-1,2-diol as an acetal chiral auxiliary afforded enol ethers (2a-f and 5a-f) of 92->95% de in 31-70% yields. Removal of the cyclohexane-1,2-diol with BF(3)-OEt(2) afforded beta-keto esters (3 and 6) bearing
Roosmarijn E Vandenbroucke et al.
Nucleic acids research, 35(12), e86-e86 (2007-06-23)
One of the major obstacles in non-viral gene transfer is the nuclear membrane. Attempts to improve the transport of DNA to the nucleus through the use of nuclear localization signals or importin-beta have achieved limited success. It has been proposed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service