Skip to Content
Merck
All Photos(1)

Key Documents

139505

Sigma-Aldrich

2-(2-Aminoethyl)-1-methylpyrrolidine

97%

Synonym(s):

1-Methyl-2-(2-aminoethyl)pyrrolidine, 1-Methyl-2-pyrrolidineethanamine, 2-(1-Methyl-2-pyrrolidinyl)ethanamine, 2-(1-Methyl-2-pyrrolidinyl)ethylamine, 2-(1-Methylpyrrolidin-2-yl)ethan-1-amine, 2-(1-Methylpyrrolidin-2-yl)ethylamine, 2-(2-Aminoethyl)-1-methylpyrrolidine, 2-(N-Methylpyrrolidin-2-yl)ethylamine, N-Methyl-2-(2-aminoethyl)pyrrolidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H16N2
CAS Number:
Molecular Weight:
128.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.4684 (lit.)

density

0.885 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CN1CCCC1CCN

InChI

1S/C7H16N2/c1-9-6-2-3-7(9)4-5-8/h7H,2-6,8H2,1H3

InChI key

PNHGJPJOMCXSKN-UHFFFAOYSA-N

General description

2-(2-Aminoethyl)-1-methylpyrrolidine labelling acts as probe within microfluidic chip for electrochemiluminescence detection. It acts as electrogenerated chemiluminescence derivatization reagent for carboxylic acid in HPLC.

Application

2-(2-Aminoethyl)-1-methylpyrrolidine was used in dual-cloud point extraction and tertiary amine labeling for the quantification of indole-3-acetic acid and indole-3-butyric acid.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xue-Bo Yin et al.
Journal of chromatography. A, 1217(8), 1399-1406 (2009-12-29)
The low concentrations of the auxins in samples of plant tissue necessitate the use of selective and sensitive techniques for their quantification. Herein a selective and sensitive method based on dual-cloud point extraction (dCPE) and tertiary amine labeling for the
M C Stoll et al.
The American journal of emergency medicine, 32(10), 1300-1300 (2014-05-23)
Thoracic injury following a major trauma can be life threatening. Veno-venous extracorporeal membrane oxygenation (vv-ECMO) can be used as a support to mechanical ventilation when acute respiratory distress syndrome is present. We report the case of an 18-year-old male driver
Hirotoshi Morita et al.
Analytical chemistry, 74(7), 1584-1589 (2002-05-30)
2-(2-Aminoethyl)-1-methylpyrrolidine and N-(3-aminopropyl)pyrrolidine (NAPP) were found to be selective and sensitive derivatization reagents for carboxylic acid by high-performance liquid chromatography (HPLC) with electrogenerated chemiluminescence detection using tris(2,2'-bipyridine)ruthenium(II). Free fatty acids and ibuprofen were used as model compounds of carboxylic acids
D F Ranney
Journal of supramolecular structure, 5(3), 335-342 (1976-01-01)
Microtubule-disrupting alkaloids and protein fixatives were used to investigate the nature of an active process that must occur within stimulator cells in order for them to initiate a unidirectional mixed lymphocyte response (MLR). Brief treatment of the stimulator cells (SC)
Alexandre Chigaev et al.
BMC immunology, 9, 26-26 (2008-06-07)
Activation of integrins in response to inside-out signaling serves as a basis for leukocyte arrest on endothelium, and migration of immune cells. Integrin-dependent adhesion is controlled by the conformational state of the molecule (i.e. change in the affinity for the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service