Skip to Content
Merck
All Photos(1)

Key Documents

137723

Sigma-Aldrich

2,4,6-Triiodophenol

97%

Synonym(s):

Bobel 24

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
I3C6H2OH
CAS Number:
Molecular Weight:
471.80
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

powder

mp

157-159 °C (lit.)

functional group

iodo

SMILES string

Oc1c(I)cc(I)cc1I

InChI

1S/C6H3I3O/c7-3-1-4(8)6(10)5(9)2-3/h1-2,10H

InChI key

VAPDZNUFNKUROY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,4,6-Triiodophenol is iodinated disinfection byproduct formed during chlorination of sewage effluents. It is a potent thyroid hormone disrupting chemical.

Application

2,4,6-Triiodophenol was used in an in vitro assay to investigate deiodinase activity in human hepatic microsomes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tingting Gong et al.
Chemosphere, 163, 359-365 (2016-08-25)
Iodide is widely present in drinking water sources as well as wastewater effluents. Chlorination and chloramination are the most commonly used disinfection methods. During chlorination or chloramination of drinking water/wastewater effluents, iodide may be oxidized to hypoiodous acid, which may
G J Miroy et al.
Proceedings of the National Academy of Sciences of the United States of America, 93(26), 15051-15056 (1996-12-24)
Transthyretin (TTR) amyloid fibril formation is observed systemically in familial amyloid polyneuropathy and senile systemic amyloidosis and appears to be the causative agent in these diseases. Herein, we demonstrate conclusively that thyroxine (10.8 microM) inhibits TTR fibril formation efficiently in
L García-Capdevila et al.
Journal of chromatography. B, Biomedical sciences and applications, 708(1-2), 169-175 (1998-07-08)
A rapid and simple HPLC method is described for the determination of Bobel-24 (2,4,6-triiodophenol) and other iodinated derivatives in biological samples. The sample preparation was liquid-liquid extraction before injection onto the HPLC system. 2,6-Diiodo-4-methylphenol was used as internal standard. Separation
Matilde Parreño et al.
Molecular cancer therapeutics, 5(5), 1166-1175 (2006-05-30)
2,4,6-Triiodophenol (Bobel-24, AM-24) was originally described as a nonsteroid antiinflammatory molecule. We have synthesized three derivatives of Bobel-24 (Bobel-4, Bobel-16, and Bobel-30) and tested their activities as putative antileukemic agents. We have found that Bobel-24 and Bobel-16 were dual inhibitors
José Antonio Castro-Hermida et al.
Veterinary parasitology, 155(3-4), 308-313 (2008-06-27)
The efficacy of the anti-inflammatory drug Bobel-24 against experimental infection by Cryptosporidium parvum was evaluated in neonatal lambs. The animals were treated by oral administration of the drug at 50 or 500 mg/kg of body weight. The prophylactic/therapeutic treatment was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service