Skip to Content
Merck
All Photos(1)

Documents

136999

Sigma-Aldrich

6-Undecanone

97%

Synonym(s):

Diamyl ketone, Dipentyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CO(CH2)4CH3
CAS Number:
Molecular Weight:
170.29
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.4270 (lit.)

bp

228 °C (lit.)

mp

14.6 °C (lit.)

density

0.831 g/mL at 25 °C (lit.)

SMILES string

CCCCCC(=O)CCCCC

InChI

1S/C11H22O/c1-3-5-7-9-11(12)10-8-6-4-2/h3-10H2,1-2H3

InChI key

ZPQAKYPOZRXKFA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

6-Undecanone acts as solvent during the analysis of lignin pyrolysis in aerosol samples. It is an efficient membrane liquid in supported liquid membrane technique for pretreatment of plasma samples prior to capillary zone electrophoresis.

Application

6-Undecanone was used in systematic screening of various organic solvents used as supported liquid membranes in electromembrane extraction.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Knut Fredrik Seip et al.
Analytical and bioanalytical chemistry, 406(8), 2151-2161 (2013-10-19)
The current work presents a large systematic screening of 61 possible organic solvents used as supported liquid membranes (SLM) in electromembrane extraction (EME). For each organic solvent, recovery, current across the SLM, and stability considerations have been investigated and correlated
S Pálmarsdóttir et al.
Journal of capillary electrophoresis, 2(4), 185-189 (1995-07-01)
The potential of using the supported liquid membrane (SLM) technique for pretreatment of plasma samples before analysis with capillary zone electrophoresis (CZE) has been investigated. A basic drug, bambuterol, was used as a model substance in a system, where either
Murtaza Hyder et al.
Journal of chromatography. A, 1249, 48-53 (2012-07-04)
A method based on three-phase hollow fiber liquid phase microextraction was developed and successfully applied to aerosols for the analysis of lignin pyrolysis acids such as syringic acid, vanillic acid and p-salicylic acid. Important parameters related to extraction process like
Knut Fredrik Seip et al.
Journal of chromatography. A, 1347, 1-7 (2014-05-06)
Electromembrane extraction (EME) was performed on samples containing substantial amounts of NaCl to investigate how the presence of salts affected the recovery, repeatability, and membrane current in the extraction system. A group of 17 non-polar basic drugs with various physical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service