Skip to Content
Merck
All Photos(4)

Documents

136816

Sigma-Aldrich

2-Methyl-3-buten-2-ol

98%

Synonym(s):

1,1-Dimethylallyl alcohol, 3-Hydroxy-3-methyl-1-butene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHC(CH3)2OH
CAS Number:
Molecular Weight:
86.13
Beilstein:
1698263
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

51 mmHg ( 25 °C)

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.416 (lit.)

bp

98-99 °C (lit.)

density

0.824 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)(O)C=C

InChI

1S/C5H10O/c1-4-5(2,3)6/h4,6H,1H2,2-3H3

InChI key

HNVRRHSXBLFLIG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Methyl-3-buten-2-ol is a biogenic volatile organic compound emitted by pine trees and a potential precursor of atmospheric secondary organic aerosol in forested regions. It is an important component of the pheromone of the bark beetle Ips typographus.

Application

2-Methyl-3-buten-2-ol was used to study the kinetics of the gas-phase reaction of OH radicals with 3-methyl-2-butenal.
Reported to be an important component of the pheromone of the bark beetle Ips typographus.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ernesto C Tuazon et al.
Physical chemistry chemical physics : PCCP, 7(11), 2298-2304 (2005-06-07)
Kinetics and products of the gas-phase reaction of OH radicals with 3-methyl-2-butenal [(CH3)2C=CHCHO] have been investigated at room temperature and atmospheric pressure of air. Using a relative rate method with methacrolein as the reference compound, a rate constant for the
Chemical and Engineering News, 57, 10-10 (1979)
Haofei Zhang et al.
Environmental science & technology, 46(17), 9437-9446 (2012-08-02)
2-Methyl-3-buten-2-ol (MBO) is an important biogenic volatile organic compound (BVOC) emitted by pine trees and a potential precursor of atmospheric secondary organic aerosol (SOA) in forested regions. In the present study, hydroxyl radical (OH)-initiated oxidation of MBO was examined in
Todd N Rosenstiel et al.
Plant physiology, 129(3), 1276-1284 (2002-07-13)
The biosynthesis and emission of volatile plant terpenoids, such as isoprene and methylbutenol (MBO), depend on the chloroplastic production of dimethylallyl diphosphate (DMAPP). To date, it has been difficult to study the relationship of cellular DMAPP levels to emission of
Reggie Spaulding et al.
Journal of the American Society for Mass Spectrometry, 13(5), 530-542 (2002-05-22)
In the western United States, in areas where emissions of the biogenic hydrocarbon, 2-methyl-3-buten-2-ol (MBO) are high, MBO contributes significantly to the oxidative capacity of the atmosphere. Hydroxyl radical oxidation of MBO can play an important role in forming tropospheric

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service