Skip to Content
Merck
All Photos(1)

Documents

135194

Sigma-Aldrich

Bromocyclohexane

98%

Synonym(s):

Cyclohexyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11Br
CAS Number:
Molecular Weight:
163.06
Beilstein:
1098373
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.495 (lit.)

bp

166-167 °C (lit.)

solubility

H2O: insoluble
alcohol: soluble

density

1.324 g/mL at 25 °C (lit.)

SMILES string

BrC1CCCCC1

InChI

1S/C6H11Br/c7-6-4-2-1-3-5-6/h6H,1-5H2

InChI key

AQNQQHJNRPDOQV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Axial conformer of bromocyclohexane forms inclusion complexes with 9,9′-bianthryl and exhibits a weak 1,3 diaxial Cl...H interaction. It participates in the alkylation reaction of of para-xylene in the presence of graphite as catalyst to yield hydrocarbon.

Application

Bromocyclohexane has been used to study the ionization of equatorial and axial conformational isomers of the chair-bromocyclohexane using vacuum ultraviolet mass-analyzed threshold ionization spectroscopy.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

145.4 °F - closed cup

Flash Point(C)

63 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Songhee Han et al.
The journal of physical chemistry. A, 114(37), 10005-10010 (2010-08-27)
Ionization of equatorial and axial conformational isomers of the chair-bromocyclohexane is investigated with use of the vacuum ultraviolet mass-analyzed threshold ionization (MATI) spectroscopic technique. Two distinct ionization energies of 9.8308 ± 0.0025 and 9.8409 ± 0.0025 eV are determined for
Alkylation on graphite in the absence of Lewis acids.
Sereda GA.
Tetrahedron Letters, 45(39), 7265-7267 (2004)
Shinya Hirano et al.
Chemical communications (Cambridge, England), (29)(29), 3646-3648 (2005-07-20)
The axial conformers of chloro- and bromocyclohexane were isolated in a pure state as inclusion complexes with 9,9'-bianthryl, and a 1,3 diaxial Cl...H weak interaction was discovered by X-ray analysis of the axial conformer of chlorocyclohexane.
Hanumantha Rao Vutukuri et al.
Angewandte Chemie (International ed. in English), 53(50), 13830-13834 (2014-11-05)
Particle shape is a critical parameter that plays an important role in self-assembly, for example, in designing targeted complex structures with desired properties. Over the last decades, an unprecedented range of monodisperse nanoparticle systems with control over the shape of
Marjolein N van der Linden et al.
Langmuir : the ACS journal of surfaces and colloids, 31(1), 65-75 (2014-12-24)
We studied suspensions of sterically stabilized poly(methyl methacrylate) (PMMA) particles in the solvent cyclohexyl bromide (CHB; εr = 7.92). We performed microelectrophoresis measurements on suspensions containing a single particle species and on binary mixtures, using confocal microscopy to measure the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service