Skip to Content
Merck
All Photos(1)

Key Documents

128848

Sigma-Aldrich

3,5-Dinitrosalicylic acid

98%

Synonym(s):

3,5-Dinitro-2-hydroxybenzoic acid, DNS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O2N)2C6H2-2-(OH)CO2H
CAS Number:
Molecular Weight:
228.12
Beilstein:
2220661
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

98%

form

powder

mp

168-172 °C (lit.)

functional group

carboxylic acid
nitro

SMILES string

OC(=O)c1cc(cc(c1O)[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C7H4N2O7/c10-6-4(7(11)12)1-3(8(13)14)2-5(6)9(15)16/h1-2,10H,(H,11,12)

InChI key

LWFUFLREGJMOIZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3,5-Dinitrosalicylic acid was used as a reagent for the preparation of oxazolines from amino alcohols[1] and for the spectrophotometric determination of ampicillin[2]. It was also used to measure the effects of silver nanoparticles on the membrane leakage of the reducing sugars[3].

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analytical Chemistry, 26, 2397-2397 (1993)
Ju Li et al.
Journal of nanoscience and nanotechnology, 13(10), 6806-6813 (2013-11-20)
Silver nanoparticles (AgNPs) with different sizes (5, 15 and 55 nm) were synthesized via simple method, and characterized by powder X-ray diffraction (XRD), transmission electron microscopy (TEM), energy-dispersive X-ray microanalysis (EDX) and ultraviolet-visible absorption spectroscopy (UV-Vis). The antibacterial activities of
Organometallics, 12, 3485-3485 (1993)
Mulazim Hussain Asim et al.
Journal of colloid and interface science, 531, 261-268 (2018-07-24)
The purpose of this study was to develop a novel mucoadhesive thiolated and S-protected gamma cyclodextrin (γ-CD) with an intact ring backbone to assure a prolonged residence time at specific target sites. Thiolated γ-CD was generated through bromine substitution of
Guillermo Coward-Kelly et al.
Biotechnology and bioengineering, 82(6), 745-749 (2003-04-04)
Two suggestions can be found in the literature to improve the reproducibility of the Mandels' filter paper assay: add supplemental cellobiase and increase the boiling time for color development. Here we provide data that strongly supports adding supplemental cellobiase. Adding

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service