Skip to Content
Merck
All Photos(3)

Key Documents

128759

Sigma-Aldrich

6-Hydroxypyridine-3-carboxylic acid

98%

Synonym(s):

2-Hydroxy-5-pyridinecarboxylic acid, 6-Hydroxynicotinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5NO3
CAS Number:
Molecular Weight:
139.11
Beilstein:
115991
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

>300 °C (lit.)

functional group

carboxylic acid

SMILES string

OC(=O)c1ccc(O)nc1

InChI

1S/C6H5NO3/c8-5-2-1-4(3-7-5)6(9)10/h1-3H,(H,7,8)(H,9,10)

InChI key

BLHCMGRVFXRYRN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

6-Hydroxypyridine-3-carboxylic acid (6-Hydroxynicotinic acid) reacts with Ln(2)O(3) (Ln = Nd, Sm, Eu, Gd) and oxalic acid (H(2)OX) to generate four novel lanthanide-organic coordination polymeric networks.

Application

6-Hydroxypyridine-3-carboxylic acid (6-Hydroxynicotinic acid) was used in the synthesis of molecularly imprinted polymer (MIP).
Employed in the synthesis of retinoids.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, 285-285 (1995)
Dayun Zhao et al.
Analytical and bioanalytical chemistry, 401(7), 2259-2273 (2011-08-27)
A new molecularly imprinted polymer (MIP) has been prepared on silica beads using the radical "grafting from" polymerization method for selective extraction of minor contaminant mycotoxin of patulin (PTL). After the introduction of amino groups onto the silica surface with
Cai-Ming Liu et al.
Dalton transactions (Cambridge, England : 2003), (29)(29), 5666-5672 (2010-05-08)
6-Hydroxypyridine-3-carboxylic acid (6-HOPy-3-CO(2)H) reacts with Ln(2)O(3) (Ln = Nd, Sm, Eu, Gd) and oxalic acid (H(2)OX) under hydrothermal conditions to generate four novel lanthanide-organic coordination polymeric networks [Ln(2)(1H-6-Opy-3-CO(2))(2)(OX)(2)(H(2)O)(3)] x 2.5 H(2)O (Ln = Nd, 1; Sm, 2; 1H-6-Opy-3-CO(2)(-) = 1-hydro-6-oxopyridine-3-carboxylate)
Ashish Gupta et al.
NMR in biomedicine, 18(5), 293-299 (2005-03-11)
The utility of (1)H NMR spectroscopy is suggested and demonstrated for the diagnosis of Pseudomonas aeruginosa in urinary tract infection (UTI). The specific property of P. aeruginosa of metabolizing nicotinic acid to 6-hydroxynicotinic acid (6-OHNA) is exploited. The quantity of
Li Xu et al.
Wei sheng wu xue bao = Acta microbiologica Sinica, 46(1), 63-67 (2006-04-04)
The current technology for microbial hydroxylation of nicotinic acid is a procedure of resting cells transformation. Here it was reported that non-degradation of 6-hydroxynicotinic acid during hydroxylation of nicotinic acid by resting cells was due to existing of nicotinic acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service