Skip to Content
Merck
All Photos(2)

Documents

128090

Sigma-Aldrich

3-Ethoxy-4-hydroxybenzaldehyde

ReagentPlus®, 99%

Synonym(s):

Ethylvanillin, NSC 1803, NSC 67240

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OC6H3(OH)CHO
CAS Number:
Molecular Weight:
166.17
Beilstein:
1073761
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

<0.01 mmHg ( 25 °C)

product line

ReagentPlus®

Assay

99%

mp

74-77 °C (lit.)

SMILES string

[H]C(=O)c1ccc(O)c(OCC)c1

InChI

1S/C9H10O3/c1-2-12-9-5-7(6-10)3-4-8(9)11/h3-6,11H,2H2,1H3

InChI key

CBOQJANXLMLOSS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethylvanillin (3-Ethoxy-4-hydroxybenzaldehyde) is an artificial additive for improving taste. It chemiluminesces by reacting with potassium permanganate in sulphuric or polyphosphoric acid media.

Application

Ethylvanillin (3-Ethoxy-4-hydroxybenzaldehyde) was used as an internal standard in the determination of phenolic compounds obtained from (alkaline) CuO-based oxidation of humic acid.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

293.0 °F - closed cup

Flash Point(C)

145 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diana L D Lima et al.
Talanta, 72(4), 1404-1409 (2007-06-15)
Operational parameters like migration time, temperature, voltage, composition of background electrolyte and content of organic modifier were optimized in CZE for the determination of lignin-like phenolic compounds. The applied background electrolyte buffer consisted of a Na(2)B(4)O(7), KH(2)PO(4) aqueous solution, pH
Meropi Timotheou-Potamia et al.
Talanta, 71(1), 208-212 (2007-01-15)
Vanillin, ethylvanillin and 4-hydroxy-3-methoxy-benzylalcohol have been found to chemiluminesce by the action of potassium permanganate in sulphuric or polyphosphoric acid media. Both acid media have been compared and sulphuric acid allows the sensitive determination of 0.15-10.0, 0.010-1.0 and 0.0030-0.30microg mL(-1)
David Kahoun et al.
Journal of chromatography. A, 1202(1), 19-33 (2008-07-16)
The objective of this study was the determination of 25 phenolic compounds in different mead samples (honeywines) using high performance liquid chromatography (HPLC) with coulometric-array detection and in case of hydroxymethylfurfural with UV detection. Our method was optimized in respect
Mónica Avila et al.
Electrophoresis, 30(19), 3413-3418 (2009-09-10)
A new strategy based on the fast separation of the fingerprint markers of Vanilla planifolia extracts and vanilla-related samples on microfluidic-electrochemistry chip is proposed. This methodology allowed the detection of all required markers for confirmation of common frauds in this
Gökçe Polat Yemiş et al.
Journal of food protection, 74(12), 2062-2069 (2011-12-22)
Preservatives could be part of an effective intervention strategy for the control of Cronobacter species in foods, but few compounds with the desired antimicrobial properties have been identified to date. We examined the antibacterial activity of vanillin, ethyl vanillin, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service