Skip to Content
Merck
All Photos(3)

Documents

125857

Sigma-Aldrich

2,3-Diaminopyridine

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7N3
CAS Number:
Molecular Weight:
109.13
Beilstein:
109869
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

powder

mp

110-115 °C (lit.)

solubility

H2O: soluble 50 mg/mL, opaque (brown to very dark brown)

SMILES string

Nc1cccnc1N

InChI

1S/C5H7N3/c6-4-2-1-3-8-5(4)7/h1-3H,6H2,(H2,7,8)

InChI key

ZZYXNRREDYWPLN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,3-Diaminopyridine forms the bis-condensed Schiff base with salicylaldehyde. It forms charge transfer complex with tetrachloro-p-benzoquinone and tetracyanoethylene.

Application

2,3-Diaminopyridine was used in the synthesis of 2,3-diaminopyridinium-4-hydroxybenzoate. It was used as internal standard in analysis of poly(amidoamine)dendrimers having different terminal functionalities by capillary electrophoresis. It was used as reagent in synthesis of imidazopyridines and organometallic complexes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of Organometallic Chemistry, 447, 299-299 (1993)
Synthesis and anti-bacterial/catalytic properties of Schiff bases and Schiff base metal complexes derived from 2, 3-diaminopyridine.
Jeewoth T, et al.
Synth. React. Inorg. Met.-Org. Chem. , 30(6), 1023-1038 (2000)
Hoong-Kun Fun et al.
Acta crystallographica. Section E, Structure reports online, 65(Pt 7), o1496-o1497 (2009-01-01)
In the title compound, C(5)H(8)N(3) (+)·C(7)H(5)O(3) (-), the pyridine N atom is protonated. In the 4-hydroxy-benzoate anion, the carboxyl-ate group is twisted slightly out of the benzene ring plane by an angle of 3.77 (5)°. The protonated N atom and one
Synthesis and Spectroscopic Studies of the Charge-Transfer Complexes of 2, 3-Diaminopyridine and p-Electron Acceptors.
Alqaradawi SY and Nour EM.
Spectroscopy Letters, 37(4), 337-345 (2004)
W J Coates et al.
Journal of medicinal chemistry, 36(10), 1387-1392 (1993-05-14)
The synthesis and phosphodiesterase (PDE) inhibitory profile of a series of imidazopyridines, including sulmazole and isomazole, on separated PDE isoenzymes are described. The results show that both sulmazole and isomazole are weak inhibitors of PDE III, and their inotropic activity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service