Skip to Content
Merck
All Photos(3)

Key Documents

124974

Sigma-Aldrich

2-Chlorobenzaldehyde

99%

Synonym(s):

o-Chlorobenzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4CHO
CAS Number:
Molecular Weight:
140.57
Beilstein:
385877
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39050404
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.84 (vs air)

Quality Level

vapor pressure

1.27 mmHg ( 50 °C)

Assay

99%

autoignition temp.

746 °F

refractive index

n20/D 1.566 (lit.)

bp

209-215 °C (lit.)

mp

9-11 °C (lit.)

density

1.248 g/mL at 25 °C (lit.)

SMILES string

[H]C(=O)c1ccccc1Cl

InChI

1S/C7H5ClO/c8-7-4-2-1-3-6(7)5-9/h1-5H

InChI key

FPYUJUBAXZAQNL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Chlorobenzaldehyde undergoes alkynylation with phenylacetylene in the presence of catalytic ligands and dimethylzinc at 0°C to form binaphthyl-derived amino alcohols.

Application

2-Chlorobenzaldehyde has been used in generation of small focused library of diversely functionalized dihydropyrimidine derivatives via one-pot three-component Biginelli cyclocondensation of β-ketoesters, aldehydes and thioureas.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

206.6 °F - closed cup

Flash Point(C)

97 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective alkynylation of aromatic aldehydes catalyzed by new chiral amino alcohol-based ligands.
Lu G, et al.
Tetrahedron Asymmetry, 12(15), 2147-2152 (2001)
Mathias J Jacobsen et al.
Organic letters, 13(13), 3418-3421 (2011-06-09)
A phosphine-mediated olefination of 2-alkynoates with aldehydes forming 1,3-dienes with high E-selectivity and up to 88% yield is described. Reaction conditions are optimized and reactions are demonstrated for various aryl, alkyl, and alkenyl aldehydes and for ethyl 2-alkynoates with different
E Schmid et al.
Mutagenesis, 6(4), 303-305 (1991-07-01)
The aneuploidy inducing capacity of 2-chlorobenzylidene malonitrile (CS), a chemical used as a riot control agent, and its hydrolysis products o-chlorobenzaldehyde and malonitrile was studied at various exposure conditions in V79 Chinese hamster cells. Chromosomes were counted in metaphase preparations
Doris Dallinger et al.
Nature protocols, 2(7), 1713-1721 (2007-07-21)
Here we report the generation of a small focused library of 12 diversely functionalized dihydropyrimidine (DHPM) derivatives via one-pot three-component Biginelli cyclocondensation of beta-ketoesters, aldehydes and (thio)ureas. By applying controlled microwave heating under sealed vessel conditions using a fully automated
Th Gomti Devi et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 62(4-5), 972-979 (2005-06-14)
The analysis of Raman anisotropy shift as a function of solvent concentration shows the weakening of pair interaction of the molecules due to the influence of solvent-induced perturbations. The present study deals with the effect of dielectric constant of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service