Skip to Content
Merck
All Photos(2)

Key Documents

124818

Sigma-Aldrich

Biphenyl-4,4′-disulfonyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClSO2C6H4C6H4SO2Cl
CAS Number:
Molecular Weight:
351.23
Beilstein:
2222668
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

205-208 °C (lit.)

SMILES string

ClS(=O)(=O)c1ccc(cc1)-c2ccc(cc2)S(Cl)(=O)=O

InChI

1S/C12H8Cl2O4S2/c13-19(15,16)11-5-1-9(2-6-11)10-3-7-12(8-4-10)20(14,17)18/h1-8H

InChI key

OTFAWEIFBPUXOH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Biphenyl-4,4′-disulfonyl chloride reacts with cycloinulohexaose in pyridine to form capped cyclofructan, 6A,6C-di-O-(biphenyl-4,4′-disulfonyl)-cycloinulohexaose.

Application

Biphenyl-4,4′-disulfonyl chloride was used in the synthesis of 6A,6D-diamino-6A,6D-dideoxy-β-cyclodextrin.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Capped cyclofructan. Preparation and structure determination of 6A, 6C-di-O-(biphenyl-4, 4'-disulfonyl)-cycloinulohexaose.
Atsumi M, et al.
Tetrahedron Letters, 35(31), 5661-5664 (1994)
Harold Cruz et al.
Chirality (2018-06-07)
Herein, we report the synthesis of C2 -symmetric sulfonamides as homogeneous and heterogeneous organocatalysts and their application in the enantioselective conjugate 1,4-Michael addition of carbonylic nucleophiles to β-nitrostyrene. Organocatalysts hydrogen bond to β-nitrostyrene and enamine in the transition state, mimicking
N Schaschke et al.
Chemistry & biology, 8(4), 313-327 (2001-04-28)
Human beta-tryptase is a mast cell specific trypsin-like serine protease that is thought to play a key role in the pathogenesis of diverse allergic and inflammatory disorders like asthma and psoriasis. The recently resolved crystal structure revealed that the enzymatically

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service