Skip to Content
Merck
All Photos(1)

Documents

119490

Sigma-Aldrich

2,4,6-Triisopropylbenzenesulfonyl chloride

97%

Synonym(s):

2,4,6-Tris(1-methylethyl)benzenesulfonyl chloride, Tripsyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CH]3C6H2SO2Cl
CAS Number:
Molecular Weight:
302.86
Beilstein:
1218575
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

92-94 °C (lit.)

solubility

ethanol: soluble 5%, clear, colorless to light yellow

SMILES string

CC(C)c1cc(C(C)C)c(c(c1)C(C)C)S(Cl)(=O)=O

InChI

1S/C15H23ClO2S/c1-9(2)12-7-13(10(3)4)15(19(16,17)18)14(8-12)11(5)6/h7-11H,1-6H3

InChI key

JAPYIBBSTJFDAK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,4,6-Triisopropylbenzenesulfonyl chloride can be used in the analysis of phosphonolipids in eggyolk by HPLC/MS technique.
It can also be used to synthesize:
  • Dimeric neomycin-neomycin conjugate with a flexible linker, 2,2′-(ethylenedioxy)bis(ethylamine).
  • Coupling reagents for the synthesis of oligonucleotides.
  • Sulfonates of pyrimidine nucleosides. These sulfonates can be displaced by nucleophiles in the presence of palladium-catalysts.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A General Route to 4-C-Substituted Pyrimidine Nucleosides
Ulrich Hennecke,et al.
Synthesis, 6, 929-935 (2007)
Studies on Polynucleotides. LII. 1 The Use of 2, 4, 6-Triisopropylbenzenesulfonyl Chloride for the Synthesis of Internucleotide Bonds2.
Lohrmann R and Khorana HG.
Journal of the American Chemical Society, 88(4), 829-833 (1966)
Analysis of glycerophosphonolipids in egg yolk.
Ternes W and Jaekel T.
European Food Research and Technology, 230(4), 559-570 (2010)
Sunil Kumar et al.
Journal of the American Chemical Society, 133(19), 7361-7375 (2011-04-29)
A dimeric neomycin-neomycin conjugate 3 with a flexible linker, 2,2'-(ethylenedioxy)bis(ethylamine), has been synthesized and characterized. Dimer 3 can selectively bind to AT-rich DNA duplexes with high affinity. Biophysical studies have been performed between 3 and different nucleic acids with varying
Mohammed A E Sallam et al.
Chirality, 16(5), 331-335 (2004-04-08)
Dehydrative cyclization of epimeric 4-(L-xylo- and L-lyxo-tetritol-1-yl)-2-phenyl-2H-1,2,3-triazoles afforded the anomeric alpha and beta-L-threofuranosyl analogs. The anomeric configuration of the formed anomeric C-nucleoside analogs was determined by circular dichroism and NMR spectroscopy.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service