Skip to Content
Merck
All Photos(1)

Key Documents

118303

Sigma-Aldrich

1,9-Decadiene

97%

Synonym(s):

Deca-1,9-diene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CH(CH2)6CH=CH2
CAS Number:
Molecular Weight:
138.25
Beilstein:
1697870
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39010313
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.432 (lit.)

bp

169 °C (lit.)

density

0.75 g/mL at 25 °C (lit.)

SMILES string

C=CCCCCCCC=C

InChI

1S/C10H18/c1-3-5-7-9-10-8-6-4-2/h3-4H,1-2,5-10H2

InChI key

NLDGJRWPPOSWLC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,9-Decadiene acts as comonomer and undergoes acyclic diene metathesis (ADMET) copolymerization with 1, 5-hexadiene to form random linear polybutadiene –polyoctenamer copolymers. It undergoes ADMET copolymerization with divinyltetraethoxydisiloxane to yield siloxylene–vinylene–alkenylene copolymer.

Application

1,9-Decadiene was used in synthesis and characterization of new telechelic polyoctenamers prepared by ADMET polymerization using Grubbs catalyst.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

105.8 °F - closed cup

Flash Point(C)

41 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Acyclic diene metathesis copolymerization of 1, 5-hexadiene and 1, 9-decadiene.
Wagener KB, et al.
Macromolecules, 23(24), 5155-5157 (1990)
Brent L Lee et al.
The Journal of chemical physics, 144(24), 245103-245103 (2016-07-03)
The time-resolved parallel artificial membrane permeability assay with fluorescence detection and comprehensive computer simulations are used to study the passive permeation of three aromatic dipeptides-N-acetyl-phenylalanineamide (NAFA), N-acetyltyrosineamide (NAYA), and N-acetyl-tryptophanamide (NATA) through a 1,2-dioleoyl-sn-glycero-3-phospocholine (DOPC) lipid bilayer. Measured permeation times
Synthesis of ABA triblock copolymers via acyclic diene metathesis polymerization and living polymerization of a-amino acid-N-carboxyanhydrides.
Brzezinska KR and Deming TJ.
Macromolecules, 34(13), 4348-4354 (2001)
ADMET copolymerization of divinyltetraethoxydisiloxane with 1, 9-decadiene catalyzed by Grubbs' catalyst.
Malecka E, et al.
J. Mol. Catal. A: Chem., 190(1), 27-31 (2002)
Shuzhe Wang et al.
Journal of medicinal chemistry, 64(17), 12761-12773 (2021-08-19)
Cyclic peptides have received increasing attention over the recent years as potential therapeutics for "undruggable" targets. One major obstacle is, however, their often relatively poor bioavailability. Here, we investigate the structure-permeability relationship of 24 cyclic decapeptides that share the same

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service