Skip to Content
Merck
All Photos(1)

Key Documents

116130

Sigma-Aldrich

Diethyl vinylphosphonate

97%

Synonym(s):

Vinylphosphonic acid diethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHPO(OCH2CH3)2
CAS Number:
Molecular Weight:
164.14
Beilstein:
507596
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.429 (lit.)

bp

202 °C (lit.)

density

1.068 g/mL at 25 °C (lit.)

functional group

phosphonate

storage temp.

2-8°C

SMILES string

CCOP(=O)(OCC)C=C

InChI

1S/C6H13O3P/c1-4-8-10(7,6-3)9-5-2/h6H,3-5H2,1-2H3

InChI key

DREPONDJUKIQLX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Diethyl vinylphosphonate (DEVP) can be used as a precursor for the synthesis of:
  • α, β-unsaturated phosphonates by reacting with arylboronic acids via Pd-catalyzed Mizoroki−Heck reaction.
  • 2-(arylamino)ethyl phosphonates by condensing with primary and secondary amines via the aza-Michael addition reaction.

It can be also employed as a monomer unit for the preparation of high-molecular-weight polymer, poly(diethyl vinylphosphonate) using lanthanide complexes.{18)
Diethyl vinylphosphonate has been used in the preparation of diethyl N-alkyl-2-aminoethylphosphonate.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Convenient synthesis of α, β-unsaturated phosphonates via a Mizoroki-Heck reaction of arylboronic acids with diethyl vinylphosphonate
Kabalka GW, et al.
Tetrahedron Letters, 45(24), 4685-4687 (2004)
Synthesis of 2-(arylamino) ethyl phosphonic acids via the aza-Michael addition on diethyl vinylphosphonate
Orm Nadine B, et al.
Tetrahedron, 69(1), 115-121 (2013)
Poly (vinylphosphonate) s synthesized by trivalent cyclopentadienyl lanthanide-induced group transfer polymerization
Salzinger S, et al.
Macromolecules, 44(15), 5920-5927 (2011)
Joachim E Klee et al.
Beilstein journal of organic chemistry, 5, 72-72 (2009-01-01)
Novel N-alkyl-N-(phosphonoethyl) substituted mono-, bis- and tris(meth)acrylamides 3 were synthesized by two different three-step reactions and characterized by IR, (1)H NMR and (13)C NMR spectroscopy as well as refractive index and viscosity. The phosphonoethyl substituted (meth)acrylamide monomers show improved hydrolytic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service