Skip to Content
Merck
All Photos(1)

Documents

115622

Sigma-Aldrich

4-Chlorobenzonitrile

99%

Synonym(s):

1-Chloro-4-benzonitrile, 1-Chloro-4-cyanobenzene, 4-Cyanophenyl chloride, p-Chlorobenzonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4CN
CAS Number:
Molecular Weight:
137.57
Beilstein:
1072122
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39050680
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

bp

223 °C (lit.)

mp

90-93 °C (lit.)

SMILES string

Clc1ccc(cc1)C#N

InChI

1S/C7H4ClN/c8-7-3-1-6(5-9)2-4-7/h1-4H

InChI key

GJNGXPDXRVXSEH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Chlorobenzonitrile has been used in the preparation of amides.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

226.4 °F

Flash Point(C)

108 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ondrej Kaplan et al.
Applied microbiology and biotechnology, 73(3), 567-575 (2006-10-25)
Aspergillus niger K10 cultivated on 2-cyanopyridine produced high levels of an intracellular nitrilase, which was partially purified (18.6-fold) with a 24% yield. The N-terminal amino acid sequence of the enzyme was highly homologous with that of a putative nitrilase from
Luca Quattrini et al.
Scientific reports, 9(1), 9943-9943 (2019-07-11)
Melanoma is the most serious form of skin cancer but its medication is still far from being safe and thoroughly effective. The search of novel therapeutic approaches represents therefore a health emergency to push through eagerly. In this study, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service