Skip to Content
Merck
All Photos(2)

Key Documents

112291

Sigma-Aldrich

Ethyl salicylate

99%

Synonym(s):

Ethyl 2-hydroxybenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(HO)C6H4CO2C2H5
CAS Number:
Molecular Weight:
166.17
Beilstein:
907659
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39024568
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

0.05 mmHg ( 25 °C)

Quality Level

Assay

99%

refractive index

n20/D 1.522 (lit.)

bp

234 °C (lit.)

mp

1 °C (lit.)

density

1.131 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

CCOC(=O)c1ccccc1O

InChI

1S/C9H10O3/c1-2-12-9(11)7-5-3-4-6-8(7)10/h3-6,10H,2H2,1H3

InChI key

GYCKQBWUSACYIF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Ethyl salicylate was hydrolysed to salicylic acid by the use of an extractive electrospray ionisation (EESI) device[1].

Application

Ethyl salicylate is a non-steroidal analgesic, anti-inflammatory and antirrheumatic drug. Its complexation with β-cyclodextrin was studied using reversed-phase liquid chromatography[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A J Collins et al.
Annals of the rheumatic diseases, 43(3), 411-415 (1984-06-01)
A topically applied rubifacient delivered by aerosol (Deep-Heat) was studied. After spray application to the forearms of volunteers, without massage, the erythema produced was measured by thermography and correlated with the concentration of 2 salicylate components of the mixture found
Xiao-lan Chen et al.
Guang pu xue yu guang pu fen xi = Guang pu, 24(3), 379-380 (2005-03-12)
Atherton-Todd reaction has been extensively applied to the synthesis of phosphates and phosphoroamidates. Under Atherton-Todd reaction conditions, dialkyl phosphites were probably transformed into diakyl phosphorochloridates. We have tried to use Atherton-Todd reaction to synthesize phosphate of salicylic-acid. But when the
A Lapczynski et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 45 Suppl 1, S397-S401 (2007-11-21)
A toxicological and dermatological review of ethyl salicylate when used as a fragrance ingredient is presented.
Amnon C Sintov et al.
Journal of controlled release : official journal of the Controlled Release Society, 79(1-3), 113-122 (2002-02-21)
The aim of this study was to investigate the percutaneous penetration and dermal metabolism of a new potential anti-acne prodrug--TU-2100 [bis(o-carboxyphenyl ethyl ester)nonanedioate] in guinea pigs. The fluxes of this agent through excised skin after applications of TU-2100 gels at
Mauricio Filippa et al.
Journal of pharmaceutical and biomedical analysis, 48(3), 969-973 (2008-07-25)
The complexation of methyl salicylate (MS) and ethyl salicylate (ES), non-steroidal analgesic, anti-inflammatory and antirrheumatic drugs with beta-cyclodextrin (betaCD) has been studied from thermodynamic and structural points of view. The complexation with betaCD has been investigated using reversed-phase liquid chromatography.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service