Skip to Content
Merck
All Photos(1)

Documents

11195

Sigma-Aldrich

L-Aspartic acid sodium salt monohydrate

≥99.0% (NT)

Synonym(s):

(S)-2-Aminobutanedioic acid sodium salt, Sodium L-aspartate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH2CH(NH2)CO2Na · H2O
CAS Number:
Molecular Weight:
173.10
Beilstein:
6101944
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

product name

L-Aspartic acid sodium salt monohydrate, ≥99.0% (NT)

Assay

≥99.0% (NT)

form

powder or crystals

optical activity

[α]20/D −14±1°, c = 1% in H2O

reaction suitability

reaction type: solution phase peptide synthesis

color

white

mp

~140 °C (dec.)

application(s)

peptide synthesis

SMILES string

[Na+].[H]O[H].N[C@@H](CC(O)=O)C([O-])=O

InChI

1S/C4H7NO4.Na.H2O/c5-2(4(8)9)1-3(6)7;;/h2H,1,5H2,(H,6,7)(H,8,9);;1H2/q;+1;/p-1/t2-;;/m0../s1

InChI key

PPTHNBYUFXSJPS-JIZZDEOASA-M

Looking for similar products? Visit Product Comparison Guide

General description

L-Aspartic acid is a non-essential amino acid.

Application

L-Aspartic acid sodium salt monohydrate can be used in studies that involve analyzing the diffusion coefficients and osmotic coefficients of amino acids in aqueous solutions.

Biochem/physiol Actions

Principal neurotransmitter for fast synaptic excitation.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sophie V Vo et al.
RSC medicinal chemistry, 11(4), 511-517 (2021-01-23)
The translocator protein (TSPO) is a target for the development of neuroinflammation imaging agents. Clinical translation of TSPO PET ligands, such as [11C]DPA-713, has been hampered by the presence of a common polymorphism (A147T TSPO), at which all second-generation TSPO
Graham Rykiel et al.
eLife, 9 (2020-10-21)
Cardiac pumping depends on the morphological structure of the heart, but also on its subcellular (ultrastructural) architecture, which enables cardiac contraction. In cases of congenital heart defects, localized ultrastructural disruptions that increase the risk of heart failure are only starting
Catriona Wimberley et al.
European journal of nuclear medicine and molecular imaging, 47(11), 2589-2601 (2020-03-27)
Longitudinal mouse PET imaging is becoming increasingly popular due to the large number of transgenic and disease models available but faces challenges. These challenges are related to the small size of the mouse brain and the limited spatial resolution of
Binary diffusion coefficients for aqueous solutions of l-aspartic acid and its respective monosodium salt.
Ribeiro AC
Journal of Solution Chemistry, 43(1), 83-92 (2014)
Activity of Water and Osmotic Coefficients for Two-and Three-Basic Amino Acid Ternary Solutions.
Tsurko EN
Journal of Chemical and Engineering Data, 57(11), 3123-3127 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service