Skip to Content
Merck
All Photos(2)

Key Documents

110469

Sigma-Aldrich

cis-Decahydronaphthalene

99%

Synonym(s):

cis-Decalin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18
CAS Number:
Molecular Weight:
138.25
Beilstein:
1900822
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.76 (vs air)

vapor pressure

42 mmHg ( 92 °C)

Assay

99%

autoignition temp.

482 °F

expl. lim.

0.7-4.9 %, 100 °F

refractive index

n20/D 1.481 (lit.)

bp

193 °C (lit.)

mp

−43 °C (lit.)

density

0.897 g/mL at 25 °C (lit.)

SMILES string

[H][C@]12CCCC[C@@]1([H])CCCC2

InChI

1S/C10H18/c1-2-6-10-8-4-3-7-9(10)5-1/h9-10H,1-8H2/t9-,10+

InChI key

NNBZCPXTIHJBJL-AOOOYVTPSA-N

Looking for similar products? Visit Product Comparison Guide

Application

cis-Decahydronaphthalene is used as a quantitation standard in the determination of sesquiterpanes.

Legal Information

Decalin is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Aquatic Chronic 2 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Corr. 1C

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

136.4 °F - closed cup

Flash Point(C)

58 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Chun Yang et al.
Journal of chromatography. A, 1216(20), 4475-4484 (2009-03-27)
This study presents a quantitative gas chromatography-mass spectrometry analysis of bicyclic sesquiterpanes (BSs) in numerous crude oils and refined petroleum products including light and mid-range distillate fuels, residual fuels, and lubricating oils collected from various sources. Ten commonly recognized bicyclic
Ken K Qian et al.
Journal of pharmaceutical sciences, 100(7), 2801-2815 (2011-02-22)
Spontaneous crystalline-to-amorphous phase transformation of organic or medicinal molecules in the presence of mesoporous materials has been observed, for which pathway was suggested to be via the vapor phase, that is, sublimation of the crystalline molecules followed by adsorption on
Mohit Kumar et al.
Nanoscale, 3(5), 2130-2133 (2011-03-30)
Naphthalene diimide (NDI) bolaamphiphilic molecules (1) self-assemble in water to form organic nanoparticles, which exhibit self-assembly induced preassociated excimer formation and hence an enhanced green fluorescence.
Sylvie Goncalves et al.
The Journal of organic chemistry, 76(9), 3274-3285 (2011-03-31)
An unprecedented and highly diastereoselective 6-endo-trig cyclization of 2-alkenyl-1,3-dithiolanes has been developed yielding trans-decalins, an important scaffold present in numerous di- and triterpenes. The novelty of this 6-endo-trig cyclization stands in the stepwise mechanism involving 2-alkenyl-1,3-dithiolane, acting as a novel
Lin Zhu et al.
Journal of natural products, 75(4), 567-571 (2012-03-08)
Five new decalin derivatives (1-5), together with two known compounds (6 and 7), were isolated from the ethyl acetate extract of red yeast rice. Their structures were elucidated by means of NMR and mass spectroscopic analyses. Monascusic lactone A (1)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service