We have not tested the solubility of this product. However, historical data indicates that this material is soluble in water at 10 mg/mL at 40 °C.
107751
5-Hydroxy-L-tryptophan
98% (calc. on dried substance)
Synonym(s):
(S)-5-Hydroxytryptophan, 5-Hydroxytryptophan, L-5-HTP, L-5-Hydroxytryptophan
About This Item
Recommended Products
Assay
98% (calc. on dried substance)
form
solid
optical activity
[α]22/D −30°, c = 1 in H2O
reaction suitability
reaction type: solution phase peptide synthesis
refractive index
n20/D 1.4850 (lit.)
mp
270 °C (dec.) (lit.)
density
0.902 g/mL at 25 °C (lit.)
application(s)
peptide synthesis
SMILES string
N[C@@H](Cc1c[nH]c2ccc(O)cc12)C(O)=O
InChI
1S/C11H12N2O3/c12-9(11(15)16)3-6-5-13-10-2-1-7(14)4-8(6)10/h1-2,4-5,9,13-14H,3,12H2,(H,15,16)/t9-/m0/s1
InChI key
LDCYZAJDBXYCGN-VIFPVBQESA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
What is the Solubility of 5-Hydroxy-L-tryptophan. 107751-1G
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service