Skip to Content
Merck
All Photos(1)

Documents

105465

Sigma-Aldrich

1,2-Dichlorobutane

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH(Cl)CH2Cl
CAS Number:
Molecular Weight:
127.01
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

28 mmHg ( 32 °C)

Assay

98%

autoignition temp.

529 °F

expl. lim.

2.25-9.25 %, 20 °F

refractive index

n20/D 1.445 (lit.)

bp

125 °C (lit.)

density

1.112 g/mL at 25 °C (lit.)

SMILES string

CCC(Cl)CCl

InChI

1S/C4H8Cl2/c1-2-4(6)3-5/h4H,2-3H2,1H3

InChI key

PQBOTZNYFQWRHU-UHFFFAOYSA-N

General description

1,2-Dichlorobutane causes the alkylation of benzene using aluminium chloride or liquid hydrogen fluoride as catalyst.

Application

1,2-Dichlorobutane was used to evaluate temperature-composition phase diagrams of binary mixtures composed of a haloalkane dissolved in either 1-ethyl-3-methylimidazolium ethylsulfate or 1-ethyl-3-methylimidazolium ethylsulfonate.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

80.6 °F - closed cup

Flash Point(C)

27 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The alkylation of benzene with 1, 2-dichloroalkanes.
Ransley D L.
The Journal of Organic Chemistry, 31(11), 3595-3599 (1966)
Camilla Vester Lauritsen et al.
International journal of food microbiology, 300, 14-21 (2019-04-17)
It is well established, that certain bacteria within the Brochothrix, Carnobacterium, Lactobacillus, Lactococcus, and Leuconostoc genera have an important role in the spoilage of chill stored poultry meat packaged in modified atmosphere. However, little is known about the role of
Francisco J Deive et al.
The journal of physical chemistry. B, 114(21), 7329-7337 (2010-05-08)
The temperature-composition phase diagrams of 40 binary mixtures composed of a haloalkane dissolved in either 1-ethyl-3-methylimidazolium ethylsulfate or 1-ethyl-3-methylimidazolium ethylsulfonate were measured from ambient temperature to the boiling point temperature of the solute. The coexistence curves corresponding to liquid-liquid equilibria
Zainab Rahmat et al.
SpringerPlus, 5(1), 1954-1954 (2016-12-10)
Paratyphoid fever caused by Salmonella enterica serovar Paratyphi A is becoming a serious health problem in Asian countries particularly Pakistan, China and India and situation is aggravated by current unavailability of a licensed vaccine. This study was designed to purify
Ali Calik et al.
Poultry science, 94(9), 2173-2182 (2015-07-19)
This study investigated the effects of dietary lactulose supplementation on broiler growth performance, intestinal histomorphology, cecal microflora, and cecal short-chain fatty acid (SCFA) concentrations. A total of 245 one-day-old male broiler chickens were randomly assigned to 5 different treatments, with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service