Skip to Content
Merck
All Photos(2)

Key Documents

103241

Sigma-Aldrich

4-Methoxy-1-naphthaldehyde

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC10H6CHO
CAS Number:
Molecular Weight:
186.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

99%

form

solid

bp

212 °C/40 mmHg (lit.)

mp

35-36 °C (lit.)

SMILES string

COc1ccc(C=O)c2ccccc12

InChI

1S/C12H10O2/c1-14-12-7-6-9(8-13)10-4-2-3-5-11(10)12/h2-8H,1H3

InChI key

MVXMNHYVCLMLDD-UHFFFAOYSA-N

Application

4-Methoxy-1-naphthaldehyde was used in fluorometric enzymatic assay for determination of activity of class I and II alcohol dehydrogenase isoenzymes in human pancreas[1] and in human liver homogenates[2].

Biochem/physiol Actions

4-Methoxy-1-naphthaldehyde is fluorogenic substrate for human alcohol dehydrogenase (ADH)[3].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lech Chrostek et al.
Journal of clinical laboratory analysis, 17(3), 93-96 (2003-04-16)
Alcohol dehydrogenase (ADH) and aldehyde dehydrogenase (ALDH), which are most abundant in the liver, are the main enzymes involved in ethanol metabolism in humans. Gender-related differences in total liver ADH and ALDH activity among different animal species have been observed
Jayaraman Jayabharathi et al.
Journal of fluorescence, 26(1), 307-316 (2015-11-21)
Efficient hole transport materials based on novel fused methoxynaphthyl phenanthrimidazole core structure were synthesised and characterized. Their device performances in phosphorescent organic light emitting diodes were investigated. The high thermal stability in combination with the reversible oxidation process made promising
L Chrostek et al.
Clinical chemistry and laboratory medicine, 38(5), 409-412 (2000-08-22)
We have measured the activity of class I and II alcohol dehydrogenase isoenzymes in the sera of patients with liver tumours, using class-specific fluorogenic naphthaldehydes as substrates. The activity of the tested isoenzymes was unchanged in primary tumours and significantly
J Wierzchowski et al.
Analytical biochemistry, 178(1), 57-62 (1989-04-01)
Two new fluorogenic substrates for human alcohol dehydrogenase (ADH), 4-methoxy-1-naphthaldehyde (IA) and 6-methoxy-2-napthaldehyde (IIA), are described. The 2-naphthaldehyde derivative fluoresces in aqueous media with a quantum yield of 0.22 with an emission maximum at 450 nm, but the 1-naphthaldehyde shows
Lech Chrostek et al.
Digestive diseases and sciences, 48(7), 1230-1233 (2003-07-23)
Ethanol metabolism in the pancreas occurs predominantly by way of an nonoxidative pathway to fatty acid ethyl esters but oxidative routes to acetaldehyde also may contribute to injury of pancreatic cells. Three metabolic systems are responsible for the oxidative metabolism

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service