Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

23280

Sigma-Aldrich

Chloranil

puriss., ≥99.0% (AT)

Synonym(s):

Tetrachloro-p-benzoquinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6Cl4O2
CAS Number:
Molecular Weight:
245.88
Beilstein/REAXYS Number:
393006
EC Number:
MDL number:
UNSPSC Code:
12162002

grade

puriss.

assay

≥99.0% (AT)

reaction suitability

reagent type: oxidant

sublimation residue

≤1%

mp

295-296 °C (dec.)

SMILES string

ClC1=C(Cl)C(=O)C(Cl)=C(Cl)C1=O

InChI

1S/C6Cl4O2/c7-1-2(8)6(12)4(10)3(9)5(1)11

InChI key

UGNWTBMOAKPKBL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

This product has been replaced by 232017-ALDRICH | Tetrachloro-1,4-benzoquinone 99%
Reagent for the dehydrogenation of hydroaromatic compounds[1]

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P.P. Fu et al.
Chemical Reviews, 78, 317-317 (1978)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service