Skip to Content
Merck
All Photos(1)

Key Documents

D-073

Supelco

5α-Dihydrotestosterone (DHT) solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

5α-Dihydrotestosterone solution, 5-alpha-Dihydrotestosterone, DHT, Dihydrotestosterone, 5α−DHT solution

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
£176.00

£176.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
£176.00

About This Item

Empirical Formula (Hill Notation):
C19H30O2
CAS Number:
Molecular Weight:
290.44
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

£176.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing
clinical testing

format

single component solution

storage temp.

−70°C

SMILES string

C[C@]12CC[C@H]3[C@@H](CC[C@H]4CC(=O)CC[C@]34C)[C@@H]1CC[C@@H]2O

InChI

1S/C19H30O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h12,14-17,21H,3-11H2,1-2H3/t12-,14-,15-,16-,17-,18-,19-/m0/s1

InChI key

NVKAWKQGWWIWPM-ABEVXSGRSA-N

General description

This Certified Spiking Solution® is suitable for GC/MS and LC/MS applications from clinical and diagnostic testing and endocrinology to sports testing.

5α-Dihydrotestosterone (DHT) is a steroid hormone and metabolite of the natural androgen, testosterone, formed by the action of NADPH-dependent enzyme 5α-reductase. It has been implicated in a broad range of diseases from male pattern baldness to benign prostatic hyperplasia and prostate cancer. As an anabolic steroid, DHT has also been used as a performance-enhancing drug to promote muscle growth.

Application

The Certified Reference Material (CRM) in solution can also be used as follows:
  • Simultaneous analysis of testosterone and 5α-dihydrotestosterone using a benchtop gas chromatograph-mass spectrophotometer (GC-MS) in human serum samples
  • Liquid chromatography-electrospray ionization-tandem mass spectrometric (LC/ESI-MS/MS) determination of testosterone and 5α-dihydrotestosterone following their proton affinitive derivatization using picolinic acid and its analogs
  • Determination of testosterone and 5α-dihydrotestosterone in plasma and testis samples of fathead minnow by a gas chromatography-tandem mass spectrometric (GC-MS/MS) method
  • Multi-residue screening of epitestosterone, testosterone, 5α-dihydrotestosterone, dehydroepiandrosterone, androsterone, and etiocholanolone in urine samples by high-resolution full scan-electrospray ionization-liquid chromatography (HR-FS-ESI-LC) combined with quadrupole orbitrap mass spectrometer (Q/Orbitrap MS) following requirements of World Antidoping Agency (WADA)
  • Quantitative analysis of nine hydroxy-androgens and their conjugates by a stable isotope dilution liquid chromatography-electrospray ionization-tandem mass spectrometry (SID-LC-ESI-MS/MS) based method following their derivatization using picolinic acid

Features and Benefits

  • Fully characterized under ISO/IEC 17025 and ISO 17034 accreditation
  • Accompanied with a comprehensive Certificate of Analysis (CoA) with data on stability, homogeneity, accuracy of concentration, uncertainty, and traceability
  • Rigorously tested through real-time stability studies to ensure accuracy and shelf life
  • Gravimetrically prepared using qualified precision balances to ensure minimal uncertainty
  • Flame sealed under argon into ampoules for long-term shelf life
  • Offered in a convenient, DEA-exempt format to improve laboratory efficiency

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Paula M Marchetti et al.
Annals of clinical biochemistry, 50(Pt 2), 95-107 (2013-02-23)
Dihydrotestosterone (DHT) is the most potent natural androgen in humans. There has been an increasing interest in this androgen and its role in the development of primary and secondary sexual characteristics as well as its potential roles in diseases ranging
Angel On Kei Chan et al.
Clinical chemistry, 59(5), 798-806 (2013-03-21)
5α-Reductase 2 deficiency (5ARD) is a known cause of 46,XY disorders of sex development (DSD). Traditionally, the diagnosis relies on dihydrotestosterone (DHT) measurement, but the results are often equivocal, potentially leading to misdiagnosis. We reviewed alternative approaches for diagnosis of
Julia Johansson et al.
PloS one, 8(1), e54357-e54357 (2013-01-26)
In rats with dihydrotestosterone (DHT)-induced polycystic ovary syndrome (PCOS), repeated low-frequency electrical stimulation of acupuncture needles restores whole-body insulin sensitivity measured by euglycemic hyperinsulinemic clamp. We hypothesized that electrical stimulation causing muscle contractions and manual stimulation causing needle sensation have
Jee In Kim et al.
American journal of physiology. Renal physiology, 305(1), F61-F70 (2013-05-10)
Male gender and the male hormone testosterone increase susceptibility to kidney ischemia and reperfusion (I/R) injury, which is associated with inflammatory responses. Possible involvement of histone deacetylase (HDAC) in inflammatory responses has been suggested. We investigated the gender-specific role of
Francesca Argellati et al.
Anticancer research, 33(3), 815-820 (2013-03-14)
To investigate periostin (POSTN) expression in the LNCaP cell line. Our LNCaP strain did not constitutively express the POSTN gene. Through cell transfection with a cloning vector, we developed an LNCaP cell line that stably expressed POSTN. LNCaP wild-type and

Articles

Accurate steroid analysis in blood serum using Supel™ Swift HLB DPX Tips; UTAK and NIST standards.

Questions

1–5 of 5 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. Does this DHT cross react between humans and mice? cat #: D-073-1ML

    1 answer
    1. DHT exhibits cross-reactivity between humans and mice, but biological effects and responses may differ due to species-specific differences. This product is an analytical standard and is not intended for animal studies.

      Helpful?

  4. Is this suitable for cell culture?

    1 answer
    1. This product is prepared in methanol, which would be toxic to cells. Currently, there is no cell culture tested product option available. However, product A8380, the powder form, has been used successfully in cell culture applications. Please see the link below to review this product. Note the Peer Reviewed Papers section lists a number of applicable publications.
      https://www.sigmaaldrich.com/product/sigma/a8380

      Helpful?

  5. hi, am I right in thinking that D-073 arrives in 1mL vials and they are to be stored long term at -70C. We only have -80C freezers, do you see that as a problem?

    1 answer
    1. The product COA (https://www.sigmaaldrich.com/certificates/Graphics/COfAInfo/fluka/pdf/cer/D-073_FE02172263.pdf) indicates on page 10 that it is okay to store this material for long term stability at -60C to -80C, so the indicated freezer/temperature should be fine.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service