[Skip to Content](https://www.sigmaaldrich.com#main-content)
[](https://www.sigmaaldrich.com/US/en)
Products
Cart0
USEN
Products
[Login / Register](https://www.sigmaaldrich.com/oidc-sign-in)
[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)
[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)
Cart0
[Home](https://www.sigmaaldrich.com/US/en)[Polymerase Chain Reaction Applications](https://www.sigmaaldrich.com/US/en/applications/genomics/pcr)Genomic DNA Extraction and Amplification Procedure using REDExtract-N-Amp™ Tissue PCR Kit
# Genomic DNA Extraction and Amplification Procedure using REDExtract-N-Amp™ Tissue PCR Kit
Product No. XNAT
The REDExtract-N-Amp™ Tissue PCR Kit contains all the reagents needed to rapidly extract and amplify genomic DNA from mouse tails and other animal tissues, buccal swabs, hair shafts, and saliva. The amplified DNA can be loaded directly onto an agarose gel after the PCR is completed and PCR products can be purified, if desired, for downstream applications such sequencing with the GenElute PCR Clean-up Kit ([NA1020](https://www.sigmaaldrich.com/US/en/product/sigma/na1020)).
- __Starting material__
- Mouse Tail (0.5 to 1.0 cm)
- Tissue (2 to 10 mg)
- Buccal (10 µL)
- __Speed__ – Extraction of genomic DNA for PCR in 15 minutes
- __Convenient__ – No long enzymatic digestions
- __Safe__ – No phenol/chloroform or other hazardous materials
- __Specific__ – Hot start antibody for highly specific PCR amplification of genomic DNA
## Materials
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
__Related Protocols__
- [Installation & Use of Milli-Q® SQ *2Series* Water Purification Systems](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/analytical-chemistry/wet-chemical-analysis/installation-use-of-milli-q-2series-water-purification-system)
- [Optimized PCR-based Detection of Mycoplasma](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/cell-culture-and-cell-culture-analysis/cell-counting-and-health-analysis/optimized-pcr-based-mycoplasma-detection)
- [Recombinant Selection](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/genomics/cloning-and-expression/restriction-enzyme-cloning-manual-recombinants)
- [Whole Genome Amplification: Blood Card Protocol](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/genomics/dna-and-rna-purification/blood-card)
- [Deoxyribonuclease I](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/genomics/dna-and-rna-purification/deoxyribonuclease-i)
- [Extract-N-Amp™ Tissue PCR Kit Protocol](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/genomics/dna-and-rna-purification/extract-n-amp-tissue-pcr-kit)
- [Saliva DNA Extraction & WGA Amplification](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/genomics/dna-and-rna-purification/extraction-of-dna-from-saliva)
- [DNA Extraction & WGA Amplification from Soil Samples](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/genomics/dna-and-rna-purification/extraction-of-dna-from-soil)
Top
__Sign In To Continue__
Sign In__Don't Have An Account?__Register
An unknown error has occured.
- English - EN
- Español - ES
[Learn More](https://www.sigmaaldrich.com)