Skip to Content
MilliporeSigma

582000

Anti-Telomerase (Ab-1) Rabbit pAb

liquid, Calbiochem®

Synonym(s):

Anti-TERT-1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

antibody product type

primary antibodies

clone

polyclonal

form

liquid

does not contain

preservative

species reactivity

human, mouse

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

General description

Rabbit polyclonal antibody supplied as undiluted serum. Recognizes the ~125 kDa telomerase (TERT) protein.
Recognizes the ~125 kDa telomerase (TERT) protein.
This Anti-Telomerase (Ab-1) Rabbit pAb is validated for use in Immunoblotting, Immunoprecipitation, Paraffin Sections for the detection of Telomerase (Ab-1).

Immunogen

Human
a synthetic peptide corresponding to amino acids from an internal region of the catalytic subunit of human telomerase reverse transcriptase (hTERT)

Application

Immunoblotting (1:100, see comments)

Immunoprecipitation (1:40, see comments)

Paraffin Sections (1:250-1:1000, see comments)

Physical form

Undiluted serum.

Preparation Note

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Antibody works only weakly in paraffin-embedded tissue sections. Nuclear extracts are recommended for immunoblotting and immunoprecipitation. Variables associated with assay conditions will dictate the optimal working dilution.
Fu, W., et al. 2000. J. Mol. Neurosci. 14, 3.
Kharbanda, S., et al. 2000. Curr. Biol. 10, 568.
Vaziri, H., et al. 1999. Mol. Cell. Biol. 19, 2373.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library