MilliporeSigma
Get 22% off for Pi Day through March 31.Save Now
Milli-Q® Ultrapure Water Solutions
Discover our expanded ultrapure system offer, designed for modern-day scientists
Find the Perfect Type 1 Lab Water Purification System
You now have more choice of Milli-Q® ultrapure (Type 1) systems to perfectly satisfy your laboratory needs — and constraints.
  • Choose from our state-of-the-art Milli-Q® IQ systems that offer a full Qualification program, advanced data traceability, and accurate TOC monitoring. Identified as Greener Alternative Products, these systems are certified to use less water, electricity, materials and packaging, and are completely mercury free (review the Design for Sustainability scorecards).
  • Or discover our Milli-Q® EQ systems with their intuitive touchscreens, agile Q-POD® dispensing, and space-saving installation options. These systems offer dedicated innovations for flexibility to adapt to your lab’s constraints. 
Milli-Q® Water Systems: Assured High Quality Ultrapure Water
With any Milli-Q® ultrapure water system, you can be sure that every dispense is high quality. Both Milli-Q® IQ and EQ 7 series systems contain the same powerful combination of purification media to provide ultrapure water quality consistently to the trace level. You can fully rely on these systems to deliver consistent water quality for your sensitive experiments, no matter your feed water quality. Plus, both types of systems can be connected to MyMilli-Q™ Remote Care monitoring and service capability, to assure productivity and quality.
Milli-Q® IQ 7003/05/10/15 Ultrapure & Pure Water System
The Ultimate Lab Water Solution
Exceed your expectations – from improved productivity to reduced environmental impact.
Milli-Q® IQ 7000 Ultrapure Water System
Invest in Lab Efficiency
See how the Milli-Q® IQ 7000 system can change the way you work in the lab.
Milli-Q® EQ 7008/16 Ultrapure & Pure Water System
Flexibility that Adapts to Your Lab
The authentic Milli-Q® ultrapure experience, designed for modern-day scientists.
What's the Best Ultrapure Water System for Your Lab?
  • Easy and agile Q-POD® dispensing. Dispensing ultrapure water is easy, precise, and effortless with the “thumb wheel”. Access a range of dispensing functions that are useful for day-to-day work in the laboratory. 
  • Intuitive touchscreen control. Go anywhere in just a few clicks. The interactive screen simplifies system use and data access.
  • Space-saving design. Only interact with the POD dispenser and touchscreen -- these compact systems can easily be integrated into any lab environment.
  • Improved sustainability. These compact systems and purification kits have reduced environmental impact vs. previous generation systems and cartridges. Polishing cartridges are 33% smaller and lighter, Lab Close mode saves water and energy, and packaging is greener. Plus, the new Drain Cap reduces the end-of-life impact of exhausted purification cartridges.
  • Tailored media optimizes performance. Purification cartridges can be adapted to your feed water situation, whether extra hard tap water, or Elix® or non-Elix® pure water feed.
  • Online monitoring & remote services. Securely manage your system from anywhere to support optimal productivity.
Compare Ultrapure Water System Features
Review the comparison table below to understand the similarities and differences between Milli-Q® IQ and EQ 7 series ultrapure water systems.
FeaturesMilli-Q® EQ 7000Milli-Q® EQ 7008/16Milli-Q® IQ 7000Milli-Q® IQ 7003/05/10/15
Final water quality gradeType 1 (ultrapure)
Intermediate water qualityN/AType 3 (RO)N/AType 2 (Elix®)
Feed water qualityPretreated*TapPretreated*Tap
Intermediate production flow rate (L/hr)N/A8 or 16N/A3, 5, 10 or 15
IPAK Gard® pretreatment packNoYesNoYes
IPAK Meta® polishing cartridgeYesYesYesNo
IPAK Quanta® polishing cartridgeYes
Application POD-Paks6
Touchscreen interfaceYes
MyMilli-Q™ Remote Care capableYes
Lab Close modeYes
Local network connectivity and USB portYes
System footprint++++
Flexibility of installation+++++++
Flexibility of Q-POD® dispenser+++++++
Number of Q-POD® dispensers1Up to 4
Dispensing flow rates38
Volumetric dispensing++++++
Drop-by-drop dispensingNoYes
Assisted dispensingNoYes
Data management
++++++
Dispense reports & QR codesNoYes
Oxidative UV lampMercuryMercury-free [Design for Sustainability scorecard]
TOC accuracyIn-line IndicatorA10® Monitor
Compatible with Milli-Q® IQ Element unit (Brochure)NoYes
QualificationNoYes
TOC calibration/verification  No†Yes
USP suitability testingNoYes
* By electrodeionization (EDI), deionization (DI), reverse osmosis (RO), or distillation technologies.
† Only temperature and conductivity cell calibration.
How to Choose a Type 1 Water Purification System
The selection of the proper ultrapure water system for your laboratory will depend on several parameters, such as:
  • Available feed water
  • Daily volume needs
  • Monitoring requirements
  • Expected certification levels
Contact our Lab Water Specialists to discuss the ultrapure water system that’s best adapted to your needs.
Select and configure the optimal ultrapure water system for your lab. 
Related Resources
Water Purification Systems, Water Purification Units & Dispensers
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
Sorry, an unexpected error has occurred
Response not successful: Received status code 500