TLC Explorer Service & Support
The TLC Explorer Documentation System is an advanced instrumental and software solution designed to provide a reliable TLC plate analysis. The device offers both visual check and software-based thin layer chromatographic analysis along with quantification. It can analyze one or more TLC plates with an area of up to 20 × 20 cm. In addition, the system uses three types of LED light sources— white light (visible spectrum), UV-A (wavelength of 366nm), and UV-C (wavelength of 254nm), to illuminate the plates from an overhead position in direct-light mode and identify distinct substance classes.
The TLC Explorer has been recognized by Select Science® as the “Best New Analytical Science Product of 2025”, an award nominated and voted for by scientists worldwide. This recognition highlights its role in enabling efficient and reliable TLC plate analysis.
Need assistance? Click here for the user manual guide.
TLC Explorer Updates
Save your time by downloading the free upgrades to keep the pre-programmed methods up-to-date and unlock new features to improve the system’s usability in your laboratory. Please find the latest free software listed below.
Request Information
All fields indicated by an * are required.
Related Products
Response not successful: Received status code 500
Related Resources
- Thin Layer Chromatography (TLC)
Choose from classical TLC plates with silica, aluminum, glass, and plastic backings as well as HPTLC plates, MS-Grade TLC and HPTLC plates, and PLC plates.
Related Video
A step-by-step tutorial video showing TLC Explorer Software, covering user setup, settings navigation, image capture, wavelength selection, track integration, data evaluation, annotations, track comparison, quantification, and report generation for TLC plate analysis.