MilliporeSigma
All Photos(1)
Key Documents
PNS1010
Sigma-Aldrich
Stabilyser Reagent
for stabilization of DNA, RNA, and protein in tissue extracts
Sign Into View Organizational & Contract Pricing
Select a Size
50 ML
$297.00
250 ML
$934.00
$297.00
Please contact Customer Service for Availability
Notify Me
Get notified when this item is ready to ship via email.
Select a Size
Change View
50 ML
$297.00
250 ML
$934.00
About This Item
UNSPSC Code:
41116012
NACRES:
NA.77
$297.00
Please contact Customer Service for Availability
Notify Me
Get notified when this item is ready to ship via email.
form
liquid
Quality Level
feature
for stabilization of DNA, RNA, and protein in tissue extracts
shipped in
wet ice
storage temp.
2-8°C
General description
The stability of protein and antibody structure and function is dependent on a variety of factors such as temperature, freeze/thaw cycles, proteases and pH. For some proteins and antibodies, function can be lost completely with a single freeze/thaw cycle and proteases can significantly degrade proteins and antibodies within hours even when kept at 4 °C. Stabilyser Reagent minimizes the risk to improve protein assay quality and accuracy.
Stabilyser Reagent is a proprietary formulation of detergents and solutes optimized for efficient extraction, stabilization and storage of protein, DNA, and RNA from tissue samples. The unique composition of Stabilyser enables isolation of functionally active proteins, while maintaining integrity of nucleic acids. Stabilization of proteins and RNA is achieved even after 24 hours at room temperature Stabilyser reagent is compatible with Western blotting protocols, functional protein activity assays, and nucleic acid applications like qPCR.
Use in conjunction with a homognenizer for simultaneous tissue lysis and stabilization of protein and nucleic acids in tissue samples.
Application
Prevents degradation of proteins and nucleic acid during tissue lysis and storage. Stabilize functional proteins and RNA.
Features and Benefits
  • Gentle, non-denaturing lysis buffer does not interfere with downstream applications
  • Convenient ready-to-use reagent
  • Stabilizes RNA and protein for analysis at a later date
Preparation Note
Ready-to-use reagent. In general it is recommended to start optimization with 20 μL of Stabilyser reagent per mg of tissue.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
Related Content
Cell lysis and protein extraction methods overview various techniques, from detergent solubilization to mechanical disruption, supporting research needs.
Questions
Reviews
No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service