Merck
HomeBiopharmaceutical CharacterizationBiosimilar Comparability Workflow for Intact Mass Analysis
Biosimilar Comparability Workflow for Intact Mass Analysis Using Trastuzumab as Model Protein
Geoffrey Rule1, Sundaram Palaniswamy2, Uma Sreenivasan3, Paresh Tank4, Fatima D’Souza4
1Bellefonte, PA, USA, 2Bangalore, India, 3Round Rock, TX, USA, 4ZRAS, Zelle Biotechnology Pvt. Ltd., Thane, India
Introduction
Biosimilar mAb drugs have surged in the last decade due to patent expiration of innovator drugs, offering affordable care to patients. Biosimilars, while sharing amino acid sequences with innovators, can differ structurally due to post-translational modifications (PTMs) resulting from changes in manufacturing processes. These variations, like N-glycosylation and alterations to termini, pose challenges in proving comparability.1
Thorough characterization of therapeutic mAbs is vital for safety and efficacy and establishing critical quality attributes (CQAs) are essential for both innovator and biosimilar drugs.2,3 Characterization generally involves chromatography (e.g., SEC, RP, HILIC) combined with mass spectrometry (MS).
Trastuzumab received approval for medical usage in the United States in September 1998 and in the European Union in August 2000. At present, five biosimilars have gained approval in both the European Union and the United States.4,5
This application describes the utilization of a nonreducing RPLC-MS workflow for intact mass comparison of trastuzumab innovator and biosimilar. The report includes the correlation of de-charged, observed masses with theoretical masses.
A recombinant human antibody standard, SILuTMLite SigmaMAbTM  (MSQC4), is utilized as a reference and assay control sample. Here, this is referred to simply as SigmaMAbTM  although several other SigmaMAbTM standards are also available commercially.
Workflow for Intact Mass Analysis of Trastuzumab
Antibody reduction (optional)
Icon for optional reduction process
    Chromatography
    Icon for chromatography
      Measurement and Analysis
      Icon for measurement and analysis
        A complete RPLC-MS workflow has been developed to simplify intact mass analysis of non-reduced monoclonal antibodies (mAbs) for biosimilarity assessment.
        In detail, it includes:
        • Mass spectrometer calibration
        • System suitability test utilizing a recombinant human monoclonal antibody reference
        • A generic RPLC-MS method compatible with sample separation and analysis of both non-reduced and reduced monoclonal antibodies
        Experimental - Sample and Reference Preparation and System Setup
        Instrument Calibration
        The Waters QToF Xevo G2XS mass spectrometer was calibrated in a mass range of 400 – 4000 m/z with a 10 μL/min infusion of 0.4 mg/mL of sodium iodide in water. Alternatively, calibration can be performed with a 10 μL/min infusion of 0.4 mg/mL cesium iodide, polyalanine in water prior to running the samples.
        System Suitability
        To evaluate performance of the entire workflow, an assay control (SigmaMAbTM) was prepared and analyzed along with the samples. SigmaMAbTM reference was tested to ensure system suitability.
        RPLC-MS System Setup and MS Data Analysis
        RPLC-MS System Setup
        The essential settings of the UHPLC-PDA chromatography system and the qToF mass spectrometer applied in the analysis of non-reduced antibodies are listed in Tables 1 and 2 below.
        Table 1.UHPLC-PDA settings.
        Table 1b.Gradient Table for intact mass analysis
        Table 2.qToF-MS settings
        MS Data Analysis
        Data were processed using the MaxEnt1 module within the UNIFI software to generate and analyze deconvoluted (zero charged) mass spectra. In general, a summed spectrum was created from the corresponding total ion chromatogram (TIC) of the eluted intact mAb. The summed m/z spectrum was then processed by the MaxEnt1 algorithm; detailed parameters are listed in Table 3.
        For glycoform analysis, data were processed using UNIFI software from WatersTM. Glycoforms were matched by the software. Glycoform relative abundance data were tabulated based on peak intensities of the coeluting glycoform species.
        Table 3.Deconvolution parameters.
        Results
        The analysis objective here, was to perform non-reduced RPLC-MS intact mass analysis on all submitted samples to compare the molecular weight of trastuzumab innovator and biosimilar drugs.
        Intact SigmaMAbTM was used to determine system suitability. All mAb samples were solubilized in 100 μL water to a final concentration of 1 mg/mL.
        System Suitability Test Results
        SigmaMAb reference sample (1 μL) was injected on the RPLC-MS system. Figure 1 illustrates the photodiode array (280 nm) and TIC (total ion current) traces of the non-reduced antibody, while Figure 2 displays the deconvoluted mass spectrum of the SigmaMAbTM reference. The observed intact mAb glycoforms matched the common glycoform masses of MSQC4, as listed in Table 4. The measured discrepancies between the observed masses and the theoretical values for four glycoforms are all within 0.011 % mass error.
        A UV chromatogram of non-reduced SigmaMAb reference standard, measured at 280 nm, displays a distinct peak at 4.72 minutes, with absorbance on the y-axis and retention time on the x-axis.
        A TIC chromatogram of non-reduced SigmaMAb reference standard displays a distinct peak at 4.84 minute, with total ion current on the y-axis and retention time on the x-axis.
        Figure 1. Photodiode array (280 nm, left) and TIC traces (right) of non-reduced SigmaMAbTM reference.
        Summed mass spectra for non-reduced SigmaMAb reference with intensity measured in counts on the y-axis and observed mass on the x-axis
        Deconvoluted mass spectra obtained for non-reduced SigmaMAb reference (MSQC4) with six distinct peaks at observed mass values of 146524, 146674, 146831, 146990, 147157, and 147311.
        Figure 2. MS data for non-reduced SigmaMAbTM reference (MSQC4). Left: summed spectrum; right: deconvoluted spectrum.
        Table 4.Observed and theoretical masses of non-reduced SigmaMAbTM reference glycoforms
        Non-Reduced Sample Results
        The monoclonal antibody samples were analyzed in their non-reduced form using RPLC-MS. The corresponding photodiode array (280 nm) traces, TICs, MS spectra and the deconvoluted MS spectra of trastuzumab innovator and biosimilar are shown in Figures 3 and 4. The relative quantitation results for N-glycoform distribution are shown in Figure 5. The method detects differences in the distribution of major N-glycoforms between innovator and biosimilar products. The observed masses of the non-reduced mAb correlate well with the calculated theoretical masses for all submitted samples, as shown in Table 5, and the observed mass error is 0.004% or less.
        A UV chromatogram of non-reduced trastuzumab, measured at 280 nm, exhibits a distinct peak at 4.67 minutes. The innovator is represented by a red curve, while the biosimilar is depicted by a blue curve, with absorbance on the y-axis and retention time on the x-axis.
        A TIC chromatogram of fully reduced trastuzumab, exhibits a distinct peak at 4.79 minutes. The innovator is represented by a red curve, while the biosimilar is depicted by a blue curve, with absorbance on the y-axis and retention time on the x-axis.
        Figure 3. Comparative photodiode array (280 nm, left) and TIC traces (right) of non-reduced trastuzumab. Red: Innovator; blue: Biosimilar.
        Summed up mass spectra for non-reduced trastuzumab. The innovator is represented by a red curve, while the biosimilar is depicted by a blue curve, with intensity measured in counts on the y-axis and observed mass on the x-axis.
        The deconvoluted mass spectra for non-reduced trastuzumab reveals distinct peaks at mass values of 148059, 148221, 148383, and 148546 from left to right. The innovator is depicted by a red curve, and the biosimilar by a blue curve, with intensity measured in counts on the y-axis and observed mass on the x-axis.
        Figure 4. Comparative MS data for non-reduced trastuzumab. Left: summed spectrum; right: deconvoluted spectrum. Red: Innovator; blue: Biosimilar
        A bar graph depicting the distribution of major N-glycoforms of trastuzumab by Intact RPLC-MS. The innovator is depicted in red curve, and the biosimilar in blue, with relative abundance measured on the y-axis.
        Figure 5.N-glycoform distribution of trastuzumab by Intact RPLC-MS. Red: Innovator; Blue: Biosimilar.
        Table 5.Calculated and experimental masses of non-reduced trastuzumab
        Conclusion
        Ensuring the regulatory approval of biosimilar monoclonal antibody (mAb) drug products necessitates a meticulous and effective comparability assessment to guarantee their safety and efficacy. This application note serves as an illustration of the utilization of reversed-phase liquid chromatography-mass spectrometry (RPLC-MS) at the intact level. The primary objective revolves around assessing the comparability between the original trastuzumab drug and its biosimilar counterpart. A comprehensive workflow was devised for intact mass analysis of non-reduced monoclonal antibodies, using trastuzumab as a representative mAb, and a SILuTMLite SigmaMAbTM product, as an assay control.
        A separate workflow (Application Note: Biosimilar Comparability Workflow by Middle-Up Mass Analysis Using Trastuzumab as Model Protein - Protocol for reduction and RPLC-MS analysis of a monoclonal antibody) describes an optional mAb reduction procedure which can be performed with either a partial, or full, reduction procedure. Depending on the procedure used, intra-chain disulfide bonds remain intact (partial reduction) or are broken (full reduction). Solutions of sodium iodide, or cesium iodide, were used for mass spectrometer calibration, and a system suitability test, utilizing a recombinant human monoclonal antibody reference, were developed for routine use. In addition, an RP method suitable for separation and analysis of both reduced and non-reduced mAbs was established using a wide-pore, superficially porous C4 column.
        Results obtained from the non-reduced SigmaMAbTM reference indicated minimal disparities of 0.011% mass error, or less, between observed and theoretical masses for four glycoforms. For trastuzumab, both the innovator and biosimilar samples displayed a strong concurrence between observed and theoretical non-reduced mAb masses, with an observed mass error of 0.004%, or lower.
        The experimental findings affirm the versatility of the workflow for analyzing non-reduced monoclonal antibody samples, delivering accurate results that enable distinct identification of multiple glycoforms.
        See more applications on the Biopharmaceutical Characterization page.
        Related Products
        Loading
        References
        1.
        Review and Approval. 2022 Dec 13 . [Internet]. U.S. Food and Drug Administration. Available from: https://www.fda.gov/drugs/biosimilars/review-and-approval
        2.
        Sandra K, Vandenheede I, Sandra P. 2014. Modern chromatographic and mass spectrometric techniques for protein biopharmaceutical characterization. Journal of Chromatography A. 133581-103. https://doi.org/10.1016/j.chroma.2013.11.057
        3.
        Berkowitz SA, Engen JR, Mazzeo JR, Jones GB. 2012. Analytical tools for characterizing biopharmaceuticals and the implications for biosimilars. Nat Rev Drug Discov. 11(7):527-540. https://doi.org/10.1038/nrd3746
        4.
        Biosimilars approved in the US. Gabionline.net. [Internet]. Available from: https://www.gabionline.net/biosimilars/general/biosimilars-approved-in-the-us
        5.
        Biosimilars approved in Europe. Gabionline.net. [Internet]. Available from: https://www.gabionline.net/biosimilars/general/biosimilars-approved-in-europe
        6.
        Protocol for purification, optional reduction, and SEC-MS analysis of a monoclonal antibody. [Internet]. Sigmaaldrich.com. Available from: /AU/en/technical-documents/protocol/pharmaceutical-and-biopharmaceutical-manufacturing/biologics-biosimilars-characterization/protocol-for-analysis-of-a-monoclonal-antibody