HPLC Analysis of Acrylamide and Acrylic Acid on Discovery® HS F5
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Discovery HS F5, 15 cm × 4.6 mm I.D., 5 μm particles (567516-U)
mobile phase
0.05% formic acid
flow rate
1.0 mL/min
column temp.
ambient
detector
UV, 206 nm
injection
1 μL
sample
1.0 mg/mL each in mobile phase
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany