[Skip to Content](https://www.sigmaaldrich.com#main-content)
[](https://www.sigmaaldrich.com/US/en)
Products
Cart0
USEN
Products
ProductsApplicationsServicesResourcesSupport
[Login / Register](https://www.sigmaaldrich.com/oidc-sign-in)
[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)
[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)
Cart0
[Home](https://www.sigmaaldrich.com/US/en)[Nuclear Magnetic Resonance (NMR)](https://www.sigmaaldrich.com/US/en/applications/analytical-chemistry/nuclear-magnetic-resonance)NMR Deuterated Solvent Properties Reference Chart
# NMR Deuterated Solvent Properties Reference Chart
__Section Overview__
- [Common Solvent Peak Coupling and Chemical Shift Values](https://www.sigmaaldrich.com#solvent)
- [NMR Solvents Reference Data](https://www.sigmaaldrich.com#nmr)
- [Related Materials](https://www.sigmaaldrich.com#materials)
## [](https://www.sigmaaldrich.com)Common Solvent Peak Coupling and Chemical Shift Values
Use the chart below to look up the coupling values — JHD and JCD(JCF) distance between multiplet peaks in hertz (Hz) — and chemical shift delta values — ðH(Mult)b and ðc(Mult)b in parts per million (ppm) — of NMR solvents by name or CAS number. Molecular weight, density (25 °C), melting point (mp), and boiling point (bp) are also given.
## [](https://www.sigmaaldrich.com)NMR Solvents Reference Data
NMR
SolventCAS. No.Mol.
wtDensity
(25 °C)MP
(°C)aBP
(°C)aðH
(Mult)bJHDðC
(Mult)bJCD (JCF)
Acetic
acid-d4
[151785](https://www.sigmaaldrich.com/US/en/product/aldrich/151785)
1186-52-364.081.11915-16115.511.53 (1) 178.4
(br)
2.03 (5)220.0 (7)20
Acetone-
d6
[151793](https://www.sigmaaldrich.com/US/en/product/aldrich/151793)666-52-464.120.872-93.855.5 206.0
(13)0.9
2.04 (5)2.229.8 (7)20
Aceto-
nitrile-d3
[151807](https://www.sigmaaldrich.com/US/en/product/aldrich/151807)2206-26-044.070.844-4880.7 118.2
(br)
1.93 (5)2.51.3 (7)21
Benzene-
d6
[151815](https://www.sigmaaldrich.com/US/en/product/aldrich/151815)1076-43-384.150.9506.879.17.15 (br) 128.0 (3)24
Chloro-form-d
CDCL3
[151823](https://www.sigmaaldrich.com/US/en/product/aldrich/151823)
865-49-6120.381.500-6460.97.24 (1) 77.0 (3)32
Cyclo-hexane-
d12
[151866](https://www.sigmaaldrich.com/US/en/product/aldrich/151866)1735-17-796.230.8934-780.71.38 (br) 26.4 (5)19
Deuterium
oxide
HDO
151882
7789-20-020.031.1073.8101.44.84 (DSS)
4.81 (TSP)
Dichloro-
methane-d2
DCM
[444324](https://www.sigmaaldrich.com/US/en/product/aldrich/444324)
1665-00-586.941.362-97405.32 (3)153.8 (5)27
N,N-Dimethyl-
formamide-d7
DMF
[189979](https://www.sigmaaldrich.com/US/en/product/aldrich/189979)
4472-41-780.141.030-611538.01 (br) 162.7 (3)30
2.91 (5)235.2 (7)21
2.74 (5)230.1 (7)21
Dimethyl
sulfoxide-d6
DMSO
[151874](https://www.sigmaaldrich.com/US/en/product/aldrich/151874)
2206-27-184.171.19016-191892.49 (5)1.739.5 (7)21
1,4-Dioxane-
d8
[186406](https://www.sigmaaldrich.com/US/en/product/aldrich/186406)17647-74-496.151.12910-12993.53 (m) 66.5 (5)22
Ethanol-
d6
[186414](https://www.sigmaaldrich.com/US/en/product/aldrich/186414)1516-08-152.110.892-114785.19 (1)
3.55 (br) 56.8 (5)22
1.11 (m) 17.2 (7)19
Methanol-d4
MEOD
[151947](https://www.sigmaaldrich.com/US/en/product/aldrich/151947)
811-98-336.070.888-9865.44.89 (1) 49.0 (7)21.4
3.30 (5)1.7
2-Propanol-d8
[175897](https://www.sigmaaldrich.com/US/en/product/aldrich/175897)
22739-76-068.140.890-89.5825.12 (1)
3.89 (br) 62.9 (3)21.5
Pyridine-
d5
[532975](https://www.sigmaaldrich.com/US/en/product/aldrich/532975)
7291-22-784.131.050-42114.48.71 (br) 149.9 (3)27.5
7.55 (br) 135.5 (3)24.5
7.19 (br) 123.5 (3)25
Tetra-hydro-furan-d8
THF
[184314](https://www.sigmaaldrich.com/US/en/product/aldrich/184314)
1693-74-980.160.985-10665-663.58 (br) 67.4 (5)22
1.73 (br) 25.3 (br)20.5
Toluene-d8
[434388](https://www.sigmaaldrich.com/US/en/product/aldrich/434388)2037-26-5100.190.943-93110 137.5 (1)
7.09 (m) 128.9 (3)23
7.00 (br) 128.0 (3)24
6.98 (m) 125.2 (3)24
2.09 (5)2.320.4 (7)19
Trifluoro-
acetic
acid-d
TFA
[152005](https://www.sigmaaldrich.com/US/en/product/aldrich/152005)
599-00-8115.031.493-15.47511.50 (1) 164.2 (4)(44)
116.6 (4)(283)
2,2,2-Trifluoro-
ethanol-d3
[396532](https://www.sigmaaldrich.com/US/en/product/aldrich/396532)77253-67-9103.061.415-4477-805.02 (1) 126.3 (4)(277)
3.88 (4 x 3)2 (9)61.5
(4 x 5)22 (36)
a Melting and boiling points (in °C) are those of the corresponding natural abundance compound (except for D2O) and are intended only to indicate the useful liquid range of the materials. b The multiplicity “br” indicates a broad peak without resolvable fine structure, while “m” denotes a broad peak with fine structure.
## Related Materials
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
__Related Articles__
- [NMR Chemical Shifts of Impurities](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/nuclear-magnetic-resonance/1h-nmr-and-13c-nmr-chemical-shifts-of-impurities-chart)
- [Pressure-Temperature Nomograph Tool](https://www.sigmaaldrich.com/US/en/support/calculators-and-apps/pressure-temperature-nomograph-interactive-tool)
- [NMR Reference Standards](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/calibration-qualification-and-validation/nmr-reference-standards)
- [Traceable Organic Certified Reference Materials for P-31 qNMR](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/calibration-qualification-and-validation/phosphorous-qnmr-organic-traceable-crm)
- [Certified Reference Materials for ¹⁹F NMR](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/calibration-qualification-and-validation/the-international-system-of-units)
- [ChemisTwin® Portal FAQ](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/nuclear-magnetic-resonance/chemistwin-portal-faq)
- [Isotopic Labeling for NMR Spectroscopy of Biological Solids](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/nuclear-magnetic-resonance/isotopic-labeling-for-nmr-spectroscopy)
- [Biomolecular NMR: Isotope Labeling Methods for Protein Dynamics Studies](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/protein-biology/protein-structural-analysis/biomolecular-nmr--isotope-labeling-methods)
__Related Product Categories__
- [Solvents](https://www.sigmaaldrich.com/US/en/language-masters/en/products/analytical-chemistry/analytical-chromatography/solvents)
- [Analytical chemistry](https://www.sigmaaldrich.com/US/en/language-masters/en/products/analytical-chemistry)
- [Analytical chromatography](https://www.sigmaaldrich.com/US/en/language-masters/en/products/analytical-chemistry/analytical-chromatography)
- [Lab chemicals](https://www.sigmaaldrich.com/US/en/language-masters/en/products/chemistry-and-biochemicals/lab-chemicals)
Top
__Sign In To Continue__
Sign In__Don't Have An Account?__Register
Support[Customer Support](https://www.sigmaaldrich.com/US/en/support/customer-support)[Contact Us](https://www.sigmaaldrich.com/US/en/collections/offices)[FAQ](https://maestro.my.site.com/knowledgeportal/s/)[Safety Data Sheets (SDS)](https://www.sigmaaldrich.com/US/en/documents-search?tab=sds)[Certificates (COA/COO)](https://www.sigmaaldrich.com/US/en/documents-search?tab=coa)[Quality & Regulatory](https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management)[Calculators & Apps](https://www.sigmaaldrich.com/US/en/support/calculators-and-apps)[Webinars](https://www.sigmaaldrich.com/US/en/collections/webinars)
Orders[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)[Custom Products](https://www.sigmaaldrich.com/US/en/services/custom-products)[eCommerce Solutions](https://www.sigmaaldrich.com/US/en/life-science/ecommerce/ecommerce-solutions)
Company[About Us](https://www.sigmaaldrich.com/US/en/life-science/about-us)[Responsibility](https://www.sigmaaldrich.com/US/en/life-science/ssbi)[Events](https://www.sigmaaldrich.com/US/en/collections/events)[Press Releases](https://www.sigmaaldrich.com/US/en/collections/press)[Programs](https://www.sigmaaldrich.com/US/en/life-science/partnership-programs)[Careers](https://careers.merckgroup.com/global/en)[Offices](https://www.sigmaaldrich.com/US/en/collections/offices)
Social Media
[](https://www.linkedin.com/company/merck-life-science)
[](https://x.com/Merck_lifesci)
[](https://www.facebook.com/Merck.lifescience)
[](https://www.instagram.com/mercklifescience/)
[](https://www.youtube.com/@MerckLifeScience/featured)
MilliporeSigma
__Research. Development. Production.__
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
[](https://www.sigmaaldrich.com/US/en)
[Sigma-Aldrich® Solutions](https://www.sigmaaldrich.com/US/en/life-science/sigma-aldrich)[BioReliance® Solutions](https://www.sigmaaldrich.com/US/en/life-science/bioreliance)[Millipore® Solutions](https://www.sigmaaldrich.com/US/en/life-science/millipore)[SAFC® Solutions](https://www.sigmaaldrich.com/US/en/life-science/safc)[Milli-Q® Solutions](https://www.sigmaaldrich.com/US/en/life-science/milliq)[Supelco® Solutions](https://www.sigmaaldrich.com/US/en/life-science/supelco)
© 2026 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved, including Text and Data Mining for AI training and similar technologies.
Reproduction of any materials from the site is strictly forbidden without permission.
[Site Use Terms](https://www.sigmaaldrich.com/US/en/life-science/legal/site-use-terms)|[Privacy Policy](https://www.sigmaaldrich.com/US/en/life-science/legal/privacy-statement)|[General Terms and Conditions of Sale](https://www.sigmaaldrich.com/US/en/life-science/legal/terms-and-conditions)|[Copyright Consent](https://www.sigmaaldrich.com/US/en/life-science/legal/copyright-consent) |[Site Map](https://www.sigmaaldrich.com/US/en/site-map)