[Skip to Content](https://www.sigmaaldrich.com#main-content) [![MilliporeSigma](https://www.sigmaaldrich.com/static/logos/purple/millipore_sigma.svg)](https://www.sigmaaldrich.com/US/en) Products Cart0 USEN Products ProductsApplicationsServicesResourcesSupport [Login / Register](https://www.sigmaaldrich.com/oidc-sign-in) [Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup) [Quick Order](https://www.sigmaaldrich.com/US/en/quick-order) Cart0 [Home](https://www.sigmaaldrich.com/US/en)[Mammalian Cell Culture](https://www.sigmaaldrich.com/US/en/applications/cell-culture-and-cell-culture-analysis/cell-culture-by-cell-type/mammalian-cell-culture)Heparin Versus Heparin Sulfate Proteoglycans # Heparin Versus Heparin Sulfate Proteoglycans - [Brief Summary](https://www.sigmaaldrich.com#summary) - [Nomenclature](https://www.sigmaaldrich.com#nomenclature) - [Heparin vs. Heparan Sulfate](https://www.sigmaaldrich.com#versus) - [Differences between heparin and heparan sulfate](https://www.sigmaaldrich.com#differences) - [Chemical Structure](https://www.sigmaaldrich.com#structure1 "Chemical Structure") - [Synthesis and Chemical Diversity](https://www.sigmaaldrich.com#synthesis1 "Synthesis and Chemical Diversity") - [Heparan Sulfate Proteoglycans](https://www.sigmaaldrich.com#partnersa "Heparan Sulfate Proteoglycans") - [Binding Partners](https://www.sigmaaldrich.com#binding-partners "Binding Partners") ## [](https://www.sigmaaldrich.com)__Brief Summary__ Heparan sulfate (HS) is a component of the extracellular matrix. It is a glycosaminoglycan that is covalently attached to core proteins to form proteoglycans. Its structure is amazingly diverse since during synthesis it undergoes extensive sulfation and epimerization. Heparin is distinct from HS in that it is produced primarily by mast cells, whereas, HS is produced by all cell types. Note that there is not a single HS or heparin structure. Instead, the basic polysaccharide components that make up these molecules vary depending upon the tissue and the developmental stage. ## [](https://www.sigmaaldrich.com)__Nomenclature__ Heparin ≠ Heparan sulfate Heparan sulfate = HS Heparan sulfate proteoglycan = HSPG ## [](https://www.sigmaaldrich.com)Heparin vs. Heparan Sulfate Heparin is only produced by mast cells. It functions as an anticoagulant. HS is made by almost all cell types and also has anticoagulant activity, but it is much lower than that of heparin. HS further varies from heparin in the degree of modification of the sugar residues. The table below summarizes the differences between the two molecules. ## [](https://www.sigmaaldrich.com)Differences between heparin and heparan sulfate | | | | | |-------------------------|---------------------|------------|---------| | | | Sulfate | Heparin | | Size | 10-70 kDa | 10-12 kDa | | | Sulfate per hexosamine | 0.8-1.8 | 1.8-2.4 | | | GlcN N-sulfates | 40-60% | >85% | | | IdoA content | 30-50% | >70% | | | Binding to antithrombin | 0-0.3% | ~30% | | | Site of synthesis | Virtually all cells | Mast cells | | Differences between heparin and heparan sulfate ## [](https://www.sigmaaldrich.com)__Chemical Structure__ Heparin and HS are glycosaminoglycans (GAGs), which are linear polysaccharides composed of two basic saccharides: an amino sugar and an uronic acid. The amino sugar is typically either N-acetyl-D-glucosamine (D-GlcNAc) or N-acetyl-D-galactosamine (D-GalNAc). The uronic acid is either D-glucuronic acid (D-GlcA) or L-iduronic acid (L-IdoA). These basic components are further varied by epimerization, sulfation, and deacetylation. Elongation and modification of the polysaccharides are thought to occur simultaneously during biosynthesis. Heparin and HS are based on a polysaccharide unit of: 01. GlcNAc (α1-4) 02. GlcA (β1-4) 03. GlcNAc (α1-4) 04. GlcA (β1-4) 05. GlcNAc-N-sulfate-6-sulfate α1-4 06. GlcA (β1-4) 07. GlcNAc-N-sulfate-6-sulfate α1-4 08. IdoA-2-sulfate (α1-4) 09. GlcNAc-N-sulfate (α1-4) 10. GlcA (β1-4) ![chemical structure](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/technical-documents/articles/cell-culture-and-analysis/mammalian-cell-culture/heparin-structure.jpg "chemical structure") Where: GlcNAc = N-acetylglucosamine lcA = Glucuronic acid IdoA = Iduronic acid They are typically covalently attached to proteins to form proteoglycans. ## [](https://www.sigmaaldrich.com)__Synthesis and chemical diversity__ The heparin disaccharide is synthesized by α-GlcNAc Transferase II and β-GlcA Transferase II. The sugar moities are then modified by at least four sulfotransferases (N-Sulfotransferase, 2-O-Sulfotransferase, 3-O-Sulfotransferase and 6-O-Sulfotransferase) and one epimerase, GlcA-C5-Epimerase. 3’-phosphoadenyl-5’-phosphosulfate (PAPS) donates the high energy sulfate groups that are covalently attached to the HS. These enzymes do not consistently modify the HS chain. Instead they act through regions so that the resulting HS polymers have areas with high degrees of sulfation and other areas with none. This results in HS chains with a staggering degree of heterogeneity. Furthermore, the patterns of epimerization and sulfation set the ligand binding sites thus changing the functionality of the chains. ## [](https://www.sigmaaldrich.com)__Heparan Sulfate Proteoglycans__ A proteoglycan (PG) is composed of a core protein with one or more covalently attached GAGs. PGs are stored in secretory granules, inserted into the plasma membrane or secreted into the ECM. The table below lists several well characterized HS proteoglycans (HSPG) along with their molecular weight and tissue localization. | | | | | |---------------|--------------------|-------------------|-------------------------------| | Proteoglycan | Core Protein (kDa) | Number of chains | Tissue | | Perlecan | 400 | 1-3 | Basement membrane | | Agrin | 212 | 2-3 | Basement membrane | | Syndecans 1-4 | 31-45 | 1-2 HS, 1-3 CS | Epithelial cells, fibroblasts | | Betaglycan | 110 | 1 HS, 1 CS | Fibroblasts | | Glypicans 1-5 | ~60 | 1-3 | Epithelial cells, fibroblasts | | Serglycin | 10-19 | 10-15 heparin, CS | Mast cells | Heparin sulfate proteoglycans present in the ECM ## [](https://www.sigmaaldrich.com)__Binding partners__ Heparin/HS have several well characterized binding proteins. Of these, one of the most extensively studied is the interaction between heparin and FGF2 (bFGF). A crystal structure of this interaction has been solved. The heparan sulfate proteoglycan (HSPG) perlecan sequesters and regulates the release of FGF, VEGF and other cytokines involved in neovascularization. | | | |-------------------------|---------------------------------| | Protein | Physiological effect of binding | | Antithrombin | Systemic anticoagulant | | t-Plasminogen activator | Clot dissolution | | FGF family | Mitogenesis | | HGF | Mitogenesis | | apoE | Lipoprotein clearance | | IL-8/MIP-1d | Inflammation | | Fibronectin | Cell adhesion | | Laminin | Cell adhesion | | Collagen V | Cell adhesion | Heparin and heparan sulfate binding proteins ## Materials Sorry, an unexpected error has occurred Response not successful: Received status code 500 ### Reference 1\. Verghese GM, Gutknecht MF, Caughey GH. 2006. Prostasin regulates epithelial monolayer function: cell-specific Gpld1-mediated secretion and functional role for GPI anchor. American Journal of Physiology-Cell Physiology. 291(6):C1258-C1270. [https://doi.org/10.1152/ajpcell.00637.2005](https://doi.org/10.1152/ajpcell.00637.2005) __Related Articles__ - [Extracellular Matrix Proteins and Tools for Cell Culture Optimization](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/3d-cell-culture/attachment-factors-for-cell-culture) - [Human Serum Albumin (HSA)](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/cell-culture-media-preparation/human-serum-albumin) - [β-Alanine in Cell Culture](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/cell-growth-and-maintenance/beta-alanine-cell-culture) - [L-Asparagine in Cell Culture](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/cell-growth-and-maintenance/l-asparagine-in-cell-culture) - [L-Ornithine in Cell Culture](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/cell-growth-and-maintenance/l-ornithine-cell-culture) - [L-Tyrosine in Cell Culture](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/cell-growth-and-maintenance/l-tyrosine-in-cell-culture) - [Corning® High Content Screening Microplates – Frequently Asked Questions](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/imaging-analysis-and-live-cell-imaging/corning-high-screening-microplates) - [Using Immortalized 16HBE14o- Human Bronchial Epithelial Cell Lines to Model Respiratory Lung Diseases](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/mammalian-cell-culture/16hbe-lung-epithelial-cell-lines) - [View More](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search) __Related Product Categories__ - [Hydrogels](https://www.sigmaaldrich.com/US/en/products/cell-culture-and-analysis/3d-cell-culture/hydrogels) Top __Sign In To Continue__ To continue reading please sign in or create an account. Sign In__Don't Have An Account?__Register An unknown error has occured.