Skip to Content
MilliporeSigma

SBP4997

OCC-1 (AB2) Blocking Peptide for PRS4997

Sign In to View Organizational & Contract Pricing.

Select a Size

0.05 MG

$78.30

$78.30


Please contact Customer Service for Availability


About This Item

NACRES:
NA.41
UNSPSC Code:
12352203

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

synthetic

form

liquid

species reactivity

human

concentration

200 μg/mL

shipped in

wet ice

storage temp.

−20°C

Quality Level

Application

Used as a blocking peptide in immunoblotting applications.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Immunogen

16 amino acids near the amino terminus of human OCC-1

Other Notes

This peptide blocks the action of antibody PRS4997.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service