Sign In to View Organizational & Contract Pricing.
Select a Size
0.05 MG
$78.30
$78.30
Please contact Customer Service for Availability
About This Item
NACRES:
NA.41
UNSPSC Code:
12352203
Skip To
biological source
synthetic
form
liquid
species reactivity
human
concentration
200 μg/mL
shipped in
wet ice
storage temp.
−20°C
Quality Level
Application
Used as a blocking peptide in immunoblotting applications.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Immunogen
7 amino acids near the carboxy terminus of human OCC-1
Other Notes
This peptide blocks the action of antibody PRS4987.
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service