Skip to Content
MilliporeSigma

HPA010551

Anti-NAXD antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CARKD, Anti-FLJ10769, Anti-LP3298, Anti-RP11-90L1.3 antibody produced in rabbit, Anti-YM014_HUMAN Isoform 2 of Q8IW45 - Homo sapiens (human)


Sign In to View Organizational & Contract Pricing.

Select a Size

100 μL

$667.00

$667.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Human Protein Atlas Number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Anti-NAXD antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

PNAVHEVEKWLPRLHALVVGPGLGRDDALLRNVQGILEVSKARDIPVVIDADGLWLVAQQPALIHGYRKAVLTPNHVEFSRLYDAVLRGPMDSDDSHGSVLRLSQALGNVTVVQ

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Quality Level

Gene Information

human ... CARKD(55739)

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
HPA012604HPA001204HPA026773
biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

product line

Prestige Antibodies® Powered by Atlas Antibodies

product line

Prestige Antibodies® Powered by Atlas Antibodies

product line

Prestige Antibodies® Powered by Atlas Antibodies

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

form

buffered aqueous glycerol solution

form

buffered aqueous glycerol solution

form

buffered aqueous glycerol solution

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CARKD (carbohydrate kinase domain containing) is responsible for dehydrating the hydrating form of NAD(P)H, thus, it is also known as NAD(P)HX.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

General description

CARKD (carbohydrate kinase domain containing) is one of the initially defined proofreading enzymes involved in intermediary metabolism. It is localized to nucleus, mitochondria and cytosol. It is an ATP-dependent epimerase and dehydratase. CARKD gene encodes which contain a mitochondrial propeptide (mCARKD), a leader peptide (spCARKD), and one which contains neither (cCARKD). spCARKD is N-glycosylated.

Immunogen

NAD(P)HX dehydratase recombinant protein epitope signature tag (PrEST)

Other Notes

Corresponding Antigen APREST71501

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexandre Y Marbaix et al.
The Journal of biological chemistry, 286(48), 41246-41252 (2011-10-14)
The reduced forms of NAD and NADP, two major nucleotides playing a central role in metabolism, are continuously damaged by enzymatic or heat-dependent hydration. We report the molecular identification of the eukaryotic dehydratase that repairs these nucleotides and show that
Alexandre Y Marbaix et al.
The Biochemical journal, 460(1), 49-58 (2014-03-13)
Hydration of NAD(P)H to NAD(P)HX, which inhibits several dehydrogenases, is corrected by an ATP-dependent dehydratase and an epimerase recently identified as the products of the vertebrate Carkd (carbohydrate kinase domain) and Aibp (apolipoprotein AI-binding protein) genes respectively. The purpose of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service