Skip to Content
MilliporeSigma
All Photos(1)

Documents

HPA007005

Sigma-Aldrich

Anti-BOLA1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-BolA-like protein 1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

SPEVLELRNESGGHAVPPGSETHFRVAVVSSRFEGLSPLQRHRLVHAALAEELGGPVHALAIQARTPAQWRENSQLDTSPPCLGGNKKTLGT

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BOLA1(51027)

Immunogen

BolA-like protein 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

BOLA1 (BolA family member 1) is an aerobic, mitochondrial protein belonging to the BolA family. It consists of a specific helix-turn-helix motif. There are three types of secreted BOLA proteins present in human, BOLA1, BOLA2, and BOLA3. It mainly prevents the changes in mitochondrial morphology caused by glutathione (GSH) depletion through balancing mitochondrial thiol-redox potential. It has also been reported that BOLA1 may also play an important role in the cell proliferation and cell-cycle regulation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70989

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter Willems et al.
Antioxidants & redox signaling, 18(2), 129-138 (2012-07-04)
The BolA protein family is widespread among eukaryotes and bacteria. In Escherichia coli, BolA causes a spherical cell shape and is overexpressed during oxidative stress. Here we aim to elucidate the possible role of its human homolog BOLA1 in mitochondrial
Takuma Kasai et al.
Protein science : a publication of the Protein Society, 13(2), 545-548 (2004-01-14)
The BolA-like proteins are widely conserved from prokaryotes to eukaryotes. The BolA-like proteins seem to be involved in cell proliferation or cell-cycle regulation, but the molecular function is still unknown. Here we determined the structure of a mouse BolA-like protein.
Yu-Bo Zhou et al.
Molecular and cellular biochemistry, 317(1-2), 61-68 (2008-06-13)
This study reported that all three human BolA proteins (hBolA1, hBolA2, and hBolA3) are novel non-classical secreted proteins identified with bioinformatics and molecular biology experiments. The three BolA fusion proteins with c-Myc tag could be secreted into the culture medium

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service