Skip to Content
MilliporeSigma

CWA-1059

Anti-Mouse CD45 (30-F11) ColorWheel® Dye-Ready mAb

greener alternative

for use with ColorWheel® Dyes (Required, sold separately)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
30-F11, monoclonal
Application:
FACS
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rat

Quality Segment

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

30-F11, monoclonal

product line

ColorWheel®

form

lyophilized

mol wt

calculated mol wt 144.84 kDa

species reactivity

mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

flow cytometry: suitable

isotype

IgG2aκ

epitope sequence

Extracellular domain

Protein ID accession no.

UniProt accession no.

compatibility

for use with ColorWheel® Dyes (Required, sold separately)

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

General description

ColorWheel® technology is a novel and proprietary method of creating your own antibody and dye combinations for use in flow cytometry. By incubating any ColorWheel® antibody with any ColorWheel® dye, researchers can quickly and simply produce primary conjugated antibodies for use in single color or multicolor flow cytometry analysis. Each ColorWheel® antibody and dye is lyophilized for long long-term storage, allowing for simplicity and flexibility without compromise. ColorWheel® technology requires both ColorWheel® antibody and ColorWheel® dye for Flow Cytometry application.
Receptor-type tyrosine-protein phosphatase C (UniProt: P06800; also known as EC:3.1.3.48, Leukocyte common antigen, L-CA, Lymphocyte antigen 5, Ly-5,T200, CD45) is encoded by the Ptprc (also known as Ly-5) gene (Gene ID: 19264) in murine species. CD45 is a single-pass type I membrane glycoprotein that is synthesized with a signal peptide (aa 1-25), which is subsequently cleaved off to generate the mature form that contains an extracellular domain (aa 26-566), a transmembrane domain (aa 567-588), and a cytoplasmic domain (aa 589-1293). It serves as a receptor-tyrosine phosphatase that is required for antigen-induced signaling in T-cells and B-cells. It contains two protein tyrosine phosphatase (PTPase) domains (aa 642-901 and 933-1216) and the first PTPase domain has enzymatic activity, while the second one seems to affect the substrate specificity of the first one. Its substrate binding site is localized to amino acids 842-848. CD45 functions as a signaling gatekeeper in T-cells that require CD45 to detect and respond to antigen because it dephosphorylates the C-terminal inhibitory tyrosines thereby activates Src family kinase Lck that phosphorylates the T cell antigen receptor (TCR). Upon T-cell activation, CD45 also it recruits and dephosphorylates SKAP1 and Fyn. Furthermore, it also dephosphorylated Lyn and modulates its activity. (Ref.: Courtney, AH., et al. (2019). Sci. Sig. 12(604); eaaw8151; Coughlin, S., et al. (2015). Proc. Natl. Acad. Sci. USA. 112(47); E6515-E6524).
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This product is Preservative-free, lyophilized product for enhanced stability and allow for ambient shipping and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.

Immunogen

Mouse thymus/spleen cells.

Application

Quality Control Testing:

Evaluated by Flow Cytometry in mouse splenocytes.

Staining of one million mouse splenocytes was performed using 5 μL of a 1:1 mixture of Cat. No. CWA-1059, Anti-Mouse CD45 (30-F11) ColorWheel® Dye-Ready mAb and Cat. No. CWD-PE ColorWheel® Antibody-Ready Phycoerythrin (PE) Dye or an equivalent amount of PE-conjugated Rat IgG2b isotype control. 

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Biochem/physiol Actions

Clone 30-F11 is a rat monoclonal antibody that detects murine CD45. It targets an epitope within the extracellular domain.

Physical form

Lyophilized from PBS containing D-Mannitol and Sucrose, normal appearance is a dried pellet. Reconstituted antibody solution is stable and functional as assessed by functional testing. Contains no biocide or preservatives, such as azide.

Preparation Note

1.0 mg/mL after reconstitution at 25 μL in PBS. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
Recommend storage of lyophilized product at 2-8°C. Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial. Reconstitute each vial by adding 25 μL of PBS. Protect from light.

Legal Information

ColorWheel is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library