Skip to Content
MilliporeSigma

CWA-1021

Anti-Human CD25 (BC96) ColorWheel® Dye-Ready mAb

greener alternative

for use with ColorWheel® Dyes (Required, sold separately)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
BC96, monoclonal
Application:
FACS
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

BC96, monoclonal

product line

ColorWheel®

form

lyophilized

mol wt

calculated mol wt 30.82 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

flow cytometry: suitable

isotype

IgG1κ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

compatibility

for use with ColorWheel® Dyes (Required, sold separately)

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

mouse ... IL2RA(3559)

General description

ColorWheel® technology is a novel and proprietary method of creating your own antibody and dye combinations for use in flow cytometry. By incubating any ColorWheel® antibody with any ColorWheel® dye, researchers can quickly and simply produce primary conjugated antibodies for use in single color or multicolor flow cytometry analysis. Each ColorWheel® antibody and dye is lyophilized for long long-term storage, allowing for simplicity and flexibility without compromise. ColorWheel® technology requires both ColorWheel® antibody and ColorWheel® dye for Flow Cytometry application.
Interleukin-2 receptor subunit alpha (UniProt: P01589; also known as IL-2 receptor subunit alpha, IL-2-RA, IL-2R subunit alpha, IL2-RA, TAC antigen, p55, CD25) is encoded by the IL2RA gene (Gene ID: 3559) in human. The IL-2 receptor (IL-2R) is composed of three different subunit complexes, the - (CD25), β- (CD122) and -chains (CD132). By itself, CD25 is not sufficient to trigger the IL-2 signaling pathway because it is a low-affinity form of the IL-2 receptor (KD = 10-8 M) that is non-functional with respect to IL-2 internalization and signaling due to its short cytoplasmic tail. It is mainly involved in IL-2 binding thereby stabilizing a secondary binding site for the β-chain and recruiting -chain to the cell surface. CD25 is a single-pass type I membrane glycoprotein that is synthesized with a signal peptide (aa 1-21), which is subsequently cleaved off to generate the mature form that contains an extracellular domain (aa 22-240), a transmembrane domain (aa 241-259), and a cytoplasmic domain (aa 260-272). CD25 is mainly expressed on activated T -cells where it serves as a receptor for Interleukin-2 and is involved in the regulation of immune tolerance by controlling regulatory T cells (TREGs) activity that suppresses the activation and expansion of autoreactive T-cells. Mutations in IL2RA gene have been linked to the development of insulin-dependent Diabetes mellitus. (Ref.: Driesen, J., et al. (2008). Immunobiology. 213(9-10); 849-858).
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This product is Preservative-free, lyophilized product for enhanced stability and allow for ambient shipping and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.

Immunogen

Human Interleukin-2 receptor a (CD25).

Application

Quality Control Testing:

Evaluated by Flow Cytometry in PHA-stimulated human peripheral blood mononuclear cells (PBMC).

Flow Cytometry Analysis: Staining of one million 3-day phytohemagglutinin (PHA)-stimulated Human peripheral blood mononuclear cells (PBMC) was performed using 5 μL of a 1:1 mixture of Cat. No. CWA-1021, Anti-Human CD25 (BC96) ColorWheel® Dye-Ready mAb or an equivalent amount of PE-conjugated Mouse IgG1 isotype control.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Biochem/physiol Actions

Clone BC96 is a mouse monoclonal antibody that detects human CD25 (Interleukin-2 receptor subunit a).

Physical form

Lyophilized from PBS containing D-Mannitol and Sucrose, normal appearance is a dried pellet. Reconstituted antibody solution is stable and functional as assessed by functional testing. Contains no biocide or preservatives, such as azide.

Preparation Note

1.0 mg/mL after reconstitution at 25 μL in PBS. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
Recommend storage of lyophilized product at 2-8°C. Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial. Reconstitute each vial by adding 25 μL of PBS. Protect from light.

Legal Information

ColorWheel is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library