Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

60350

Sigma-Aldrich

Potassium hydrogen diiodate

puriss. p.a., ≥99.8% (RT)

Synonym(s):

Potassium biiodate, Potassium hydrogeniodate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KH(IO3)2
CAS Number:
Molecular Weight:
389.91
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21

grade

puriss. p.a.

Assay

≥99.8% (RT)

form

powder or crystals

impurities

≤0.002% total nitrogen (N)

pH

~1-2 (20 °C, 50 g/L)

anion traces

bromide, bromate, chloride, chlorate (as Cl-): ≤200 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤20 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤20 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

[K+].OI(=O)=O.[O-]I(=O)=O

InChI

1S/2HIO3.K/c2*2-1(3)4;/h2*(H,2,3,4);/q;;+1/p-1

InChI key

ACAYDTMSDROWHW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Ox. Sol. 2 - Skin Corr. 1B

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Allen E Haddrell et al.
The journal of physical chemistry. A, 116(40), 9941-9953 (2012-09-18)
The most used instrument in single particle hygroscopic analysis over the past thirty years has been the electrodynamic balance (EDB). Two general assumptions are made in hygroscopic studies involving the EDB. First, it is assumed that the net charge on
Barbara Krassowska-Swiebocka et al.
Molecules (Basel, Switzerland), 10(2), 394-400 (2007-11-17)
Benzene, halobenzenes, and a number of more or less deactivated arenes,including nitrobenzene, readily reacted in anhydrous HIO3/AcOH/Ac2O/conc. H2SO4 mixtures to probably give ArIO2 intermediates or other hypervalent species (not isolated). The final reaction mixtures were poured into excess aq. Na2SO3
[Correlation between ERPF and retention function on 131I-OIH renogram].
M Maeda et al.
Kaku igaku. The Japanese journal of nuclear medicine, 21(10), 1285-1292 (1984-10-01)
K J Hill et al.
Biochimica et biophysica acta, 1326(1), 37-46 (1997-05-22)
Liposomes were prepared from phospholipid mixtures of dipalmitoylphosphatidylcholine (DPPC) and phosphatidylinositol (PI), encapsulating the enzymes glucose oxidase (GO) and GO in combination with horse radish peroxidase (HRP) by both extrusion (VET) and reverse-phase evaporation (REV). The optimum level of PI
Colour reactions of iodic acid as reagent for identifying drugs by thin-layer chromatography.
G Cavazzutti et al.
Journal of chromatography, 268(3), 528-534 (1983-10-07)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service