MilliporeSigma
Key Documents
Skip To
807796
Cyrene™
BioRenewable, DMF and NMP Substitute
Promotion Available
Sustainability features
Synonym(s):
Dihydrolevoglucosenone
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Size/SKUAvailabilityPrice
100 mL
Available to ship TODAY
$145.00
1 L
Available to ship TODAYfromMILWAUKEE
$268.00
2.5 L
Available to ship TODAYfromMILWAUKEE
$527.00
4 L
Available to ship TODAYfromMILWAUKEE
$717.00
18 L
Available to ship TODAYfromMILWAUKEE
$2,500.00
About This Item
Empirical Formula (Hill Notation):
C6H8O3
CAS Number:
Molecular Weight:
128.13
MDL number:
UNSPSC Code:
12191600
NACRES:
NA.21
Assay:
≥98.5% (GC)
Technique(s):
: 0.5% using Water (by Karl Fischer)
Bp:
227 °C
$145.00
Available to ship TODAYDetails
​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Quality Segment
assay
≥98.5% (GC)
form
liquid
sustainability
Sustainability features
technique(s)
: 0.5% using Water (by Karl Fischer)
bp
227 °C
mp
-18 °C
density
1.25 g/mL
greener alternative category
SMILES string
O1[C@H]2OC[C@@H]1CCC2=O
InChI
1S/C6H8O3/c7-5-2-1-4-3-8-6(5)9-4/h4,6H,1-3H2/t4-,6+/m0/s1
InChI key
WHIRALQRTSITMI-UJURSFKZSA-N
General description
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is BioRenewable and thus aligns with "Designing Safer Chemicals", ″Safer Solvents and Auxiliaries″ and ″Use of Renewable Feedstocks″. Click here for more information.
Cyrene is a biobased dipolar, safe for end-of-life disposal, decomposing into CO2 and H2O. It is an aprotic alternative to common solvents that are of environmental concern. Cyrene™ is an alternative for many solvents classified by REACH as Substances of Very High Concern (SVHC), such as N-Methylpyrrolidone (NMP) and N,N-Dimethylformamide (DMF).
Application
Cyrene has been applied in replacement of NMP and/or DMF in some applications, such as demonstrating to exceed NMP′s dispersive ability for graphene solutions due to its optimum polarity and high viscosity, creating larger and less defective graphene flakes. Find below more uses for Cyrene™.
1. Dispersive ability for graphene solutions.
2. Alternative to DMF in the synthesis of metal-organic frameworks.
3. Organic synthesis:
Features and Benefits
  • 99% biodegradation in 28 days
  • Stable during incineration
  • Not mutagenic or genotoxic
  • ASTM D6866 - Standard Test Methods for Determining the Biobased Content
  • Made from Renewable Resource − Cellulose
Legal Information
CYRENE is a trademark of Circa Group Pvt Ltd
Compare Similar Items
View Full Comparison
Show Differences
1 of 1
This Item
92019392020760109
description
BioRenewable, DMF and NMP Substitute
description
BioRenewable, DMF and NMP Substitute
description
BioRenewable, DMF and NMP Substitute
description
puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Quality Level
100
Quality Level
-
Quality Level
-
Quality Level
200
technique(s)
: 0.5% using Water (by Karl Fischer)
technique(s)
-
technique(s)
-
technique(s)
-
assay
≥98.5% (GC)
assay
-
assay
-
assay
≥99.0% (T)
form
liquid
form
liquid
form
liquid
form
powder or crystals
bp
227 °C
bp
-
bp
-
bp
-
density
1.25 g/mL
density
1.153 g/mL
density
1.141 g/mL
density
-
Pictograms
Exclamation mark
Signal Word
Warning
Hazard Codes
Hazard Classifications
Eye Irrit. 2
Storage Class
10 - Combustible liquids
Flash Point (°F)
226.4 °F - closed cup
Flash Point (°C)
108 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
Questions
1–8 of 8 Questions  
  1. What are is the optical enantiomeric purity of this Cyrene?
    1 answer
    1. There is no enantiomeric purity specified for this product. However, by definition this material is the R enantiomer. The optical rotation for each lot is determined and reported in the Certificate of Analysis.
      Helpful?
  2. How can I determine the shelf life / expiration / retest date of this product?
    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf
      Helpful?
  3. How is shipping temperature determined? And how is it related to the product storage temperature?
    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
      Helpful?
  4. Could you provide information on whether Cyrene has any softening or solubilizing effects on ABS and PET-G plastics?
    1 answer
    1. Unfortunately, the compatibility of Cyrene (Dihydrolevoglucosenone) with materials such as ABS and PET-G has not been investigated. This compound is known to dissolve a number of polymeric materials. Please see the link below to review a publication that may be helpful:
      https://www.sciencedirect.com/science/article/pii/S2352554121002187
      Helpful?
  5. What is the recommended operating temperature for stripping photoresist on Silicon or GaAs semiconductor wafers when using Cyrene?
    1 answer
    1. It is known that research is currently underway in this area and Cyrene™ has shown promise. However, due to the confidential nature of these processes, a specific recommended operating temperature cannot be provided. Nevertheless, it's worth noting that the safe operating temperature of neat Cyrene™ is below 140 °C. It is advisable for the user to conduct additional safety checks and tests for their specific application.
      Helpful?
  6. What is a BioRenewable solvent?
    1 answer
    1. BioRenewable solvents are sourced from renewable and sustainable biobased materials, such as soy/palm oil, wood waste, hay or corn byproducts, etc. It is important to highlight that our BioRenewable products do not compete with food production.
      Helpful?
  7. Do BioRenewable solvents have a similar quality to petroleum-based solvents?
    1 answer
    1. BioRenewable solvents can have the same or higher purity level compared to traditional solvents. These solvents are free of the many byproducts of petroleum manufacturing, such as benzene, aldehydes, and ethers.
      Helpful?
  8. Why should you use BioRenewable solvents?
    1 answer
    1. You will contribute to create a sustainable world, significantly lowering environmental impact. Biorenewable solvents also decrease petroleum sources dependency, have higher recovery rates, and/or are safer for handling and disposal.
      Helpful?
Reviews
1–2 of 3 Reviews  
Active Filters
  1. Oslo
    • Review 1
    • Votes 36
    5 out of 5 stars.
    Cyrene as a polar solvent
    We have used Cyrene as a replacement solvent to DMSO and it has worked very good. It dissolves all of the reagents very well, and the resulting product is very similar. There is some odor to it, but as long as you work in a fume hood it is not a problem.
    Helpful?
    • Review 1
    • Votes 0
    5 out of 5 stars.
    Wonderful Solvent
    Works wonderfully as a substitute for NMP, DMF, or Cyclohexanone/Terpineol. Has a higher viscosity than most of them, so of course keep that in mind.
    Helpful?
  2. 1 Ratings-Only Review