[Skip to Content](https://www.sigmaaldrich.com#main-content) [![MilliporeSigma](https://www.sigmaaldrich.com/static/logos/purple/millipore_sigma.svg)](https://www.sigmaaldrich.com/US/en) Products Cart0 USEN Products [Login / Register](https://www.sigmaaldrich.com/oidc-sign-in) [Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup) [Quick Order](https://www.sigmaaldrich.com/US/en/quick-order) Cart0 [Home](https://www.sigmaaldrich.com/US/en)[Mammalian Cell Culture](https://www.sigmaaldrich.com/US/en/applications/cell-culture-and-cell-culture-analysis/cell-culture-by-cell-type/mammalian-cell-culture)Equipment for the Cell Culture Laboratory # Equipment for the Cell Culture Laboratory *ECACC Laboratory Handbook 4th Edition* ## Laboratory Design Perhaps one of the most undervalued aspects of [tissue culture](https://www.sigmaaldrich.com/US/en/products/cell-culture-and-analysis/cell-culture-media-and-buffers) is the need to design the facility to ensure that good quality material is produced in a safe and efficient manner. Most tissue culture is undertaken in laboratories that have been adapted for the purpose and in conditions that are not ideal. However, as long as a few basic guidelines are adopted this should not compromise the work. There are several aspects to the design of good tissue culture facilities. Ideally work should be conducted in a single use facility which, if at all possible, should be separated into an area reserved for handling newly received material (quarantine area) and an area for material which is known to be free of contaminants (main tissue culture facility). If this is not possible work should be separated by time with all manipulations on clean material being completed prior to manipulations involving the ‘quarantine’ material. Separate incubators should also be designated. In addition, the work surfaces should be thoroughly cleaned and disinfected between activities. ![Laboratory Design](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/technical-documents/articles/cell-culture-and-analysis/mammalian-cell-culture/laboratory-design.jpg "Laboratory Design") All new material should be handled as ‘quarantine’ material until it has been shown to be free of contaminants such as bacteria, fungi and particularly mycoplasma. Conducting tissue culture in a shared facility requires considerable planning and it is essential that a good aseptic technique is used throughout to minimize the risk of contamination. For most cell lines, the laboratory should be designated to at least BSL2/Category 2 based on the Advisory Committee on Dangerous Pathogens (ACDP) guidelines (ACDP, 1995)†. However, the precise category required is dependent upon the cell line and nature of the work proposed. The guidelines make recommendations regarding the laboratory environment including lighting, heating, the type of work surfaces and flooring and provision of handwashing facilities. In addition, it is recommended that laboratories should be run at air pressures that are negative to corridors to contain any risks within the laboratory. † (1995) Categorization of Biological Agents According to Hazard and Categories of Containment, 4th ## Cell Culture Biosafety Cabinets A biosafety cabinet, or laminar flow hood, is probably the most important piece of equipment for cell culture since, when operated correctly, it will provide a clean working environment for the product, whilst protecting the operator from aerosols. In these cabinets operator and/or product protection is provided using HEPA (high efficiency particulate air) filters. The level of containment provided varies according to the class of cabinet used. Cabinets may be ducted to atmosphere or re-circulated through a second HEPA filter before passing to atmosphere (__Figure 1__). Environmental monitoring with Tryptone Soy Broth agar settle plates inside the cabinet for a minimum of four hours is a good indicator of how clean a cabinet is. There should be no growth of bacteria after incubation for 3 days, or of fungi after incubation for 5 days, on such plates. In most cases a class 2 cabinet is adequate for cell culture. However, each study must be assessed for its hazard risk and it is possible that additional factors, such as a known virus infection or an uncertain provenance may require a higher level of containment. ![Diagram of cell culture biosafety cabinet airflow patterns](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/technical-documents/articles/cell-culture-and-analysis/mammalian-cell-culture/cc-biosafety-cabinets.jpg "Diagram of cell culture biosafety cabinet airflow patterns") __Figure 1.__ ## Cell Culture Incubators Cell cultures require a strictly controlled environment in which to grow. Cell culture incubators are used routinely to provide the correct growth conditions, such as temperature, degree of humidity and CO2 levels, in a controlled and stable manner. Generally, they can be set to run at temperatures in the range of 28 °C (for insect cell lines) to 37 °C (for mammalian cell lines) and set to provide CO2 at the required level (e.g. 5-10%). Some incubators also have the facility to control the O2 levels. Copper-coated incubators are also now available. These are reported to reduce the risk of microbial contamination within the incubator due to the microbial inhibitory activity of copper. The inclusion of a [bactericidal agent](https://www.sigmaaldrich.com/US/en/product/sigma/s5525) in the incubator water trays will also reduce the risk of bacterial and fungal growth. However, there is no substitute for regular cleaning and disinfection. ![Cell Culture Incubators](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/technical-documents/articles/cell-culture-and-analysis/mammalian-cell-culture/cell-culture-incubators.jpg "Cell Culture Incubators") ## Work Surfaces and Flooring In order to maintain a clean working environment, the laboratory surfaces including bench-tops, walls and flooring should be smooth and easy to clean. They should also be waterproof and resistant to a variety of chemicals (such as acids, alkalis, solvents and disinfectants). In areas used for the storage of materials in liquid nitrogen, the floors should be resistant to cracking if any liquid nitrogen is spilled. In addition, the floors and walls should be continuous with a coved skirting area to make cleaning easier and reduce the potential for dust to accumulate. Windows should be sealed. Work surfaces should be positioned at a comfortable working height. ## Cell Culture Plasticware and Consumables Almost every type of cell culture vessel, together with support consumables such as tubes and pipettes, are commercially available as single use, sterile packs. The use of such plasticware is more cost effective than recycling glassware, enables a higher level of quality assurance and removes the need for validation of cleaning and sterilization procedures. Plastic tissue culture flasks are usually either treated to provide a hydrophilic surface to facilitate attachment of anchorage dependent cells or untreated to provide a hydrophobic surface for the culture of cells in suspension. ![Cell Culture Plasticware and Consumables](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/technical-documents/articles/cell-culture-and-analysis/mammalian-cell-culture/cc-plasticware-consumables.jpg "Cell Culture Plasticware and Consumables") | | | | |---------------------|-------------------|--------------------------------------------------------------------------------------------------------------------------------------------------------------------| | Cell Culture Flasks | HYPERFlask® | [Stericup® Filters](https://www.sigmaaldrich.com/US/en/products/cell-culture-and-analysis/sterile-lab-media-filtration-and-cultureware/stericup-vacuum-filtration) | | Roller Bottles | Pipettes | Spinner Flasks | | Cryogenic Vials | Multi-Well Plates | Culture Tubes | | Cell Culture Plates | Centrifuge Tubes | Millex® Syringe Filters | ## Care and Maintenance of Laboratory Areas In order to maintain a clean and safe working environment, tidiness and cleanliness are key. All spills should be dealt with immediately. Routine cleaning should be undertaken involving the cleaning of all work surfaces both inside and outside of the microbiological safety cabinet, the floors and all other pieces of equipment (e.g. centrifuges) with [70% ethanol](https://www.sigmaaldrich.com/US/en/product/sial/793213). Additionally, the elimination of potential [mycoplasma contamination](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/cell-counting-and-health-analysis/mycoplasma-detection-and-elimination) of cells and culture media must be achieved. Humidified incubators are a particular area for concern due to the potential for fungal and bacterial growth in the water trays. This will create a contamination risk that can only be avoided by regular cleaning of the incubator. All major pieces of equipment should be regularly maintained and serviced by qualified engineers, for example: - Biosafety cabinets should be checked every six months to ensure that they are safe to use in terms of product and user protection. These tests confirm that the airflow is correct and that the HEPA filters are functioning properly. - The temperature of an incubator should be regularly checked with a NAMAS (National Accreditation of Measurement and Sampling, UK), or equivalent calibrated thermometer, and temperature adjusted as necessary. - Incubator CO2 and O2 levels should also be regularly checked to ensure the levels are being maintained correctly. __Related Articles__ - [Extracellular Matrix Proteins and Tools for Cell Culture Optimization](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/3d-cell-culture/attachment-factors-for-cell-culture) - [Human Serum Albumin (HSA)](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/cell-culture-media-preparation/human-serum-albumin) - [β-Alanine in Cell Culture](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/cell-growth-and-maintenance/beta-alanine-cell-culture) - [L-Asparagine in Cell Culture](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/cell-growth-and-maintenance/l-asparagine-in-cell-culture) - [L-Ornithine in Cell Culture](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/cell-growth-and-maintenance/l-ornithine-cell-culture) - [L-Tyrosine in Cell Culture](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/cell-growth-and-maintenance/l-tyrosine-in-cell-culture) - [Corning® High Content Screening Microplates – Frequently Asked Questions](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/imaging-analysis-and-live-cell-imaging/corning-high-screening-microplates) - [Using Immortalized 16HBE14o- Human Bronchial Epithelial Cell Lines to Model Respiratory Lung Diseases](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/mammalian-cell-culture/16hbe-lung-epithelial-cell-lines) - [View More](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search) Top __Sign In To Continue__ To continue reading please sign in or create an account. Sign In__Don't Have An Account?__Register An unknown error has occured. - English - EN - Español - ES [Learn More](https://www.sigmaaldrich.com)