Skip to Content
MilliporeSigma
2-(Diethylamino)ethanol ≥99.5%

2-(Diethylamino)ethanol

2-(Diethylamino)ethanol ≥99.5%
Synonym(s):
N,N-Diethylethanolamine, DEAE, DEEA, 2-(Diethylamino)-ethanol
Linear Formula:
(C2H5)2NCH2CH2OH
CAS Number:
Molecular Weight:
117.19

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing