Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P5420

Sigma-Aldrich

Pullulanase from Klebsiella pneumoniae

ammonium sulfate suspension, ≥5 units/mg protein (biuret)

Synonym(s):

Amylopectin 6-gluconohydrolase, Limit dextrinase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204

form

ammonium sulfate suspension

specific activity

≥5 units/mg protein (biuret)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Pullulanase is a glycolytic enzyme from Klebsiella pneumoniae that is used to hydrolyze pullulan, an α-1,6-linked homopolymer of maltotriose[1]. Product P5420 has been used to degrade carbohydrate from white beans Phaseolus vulgaris L. in healthy humans[2] and to study pullulan biosynthesis[1].
The enzyme from Sigma has been used to estimate the pullulan content of the ethanol precipitates obtained from fermented agro-industrial wastes.[3] It has also been used to the evaluate the branching of the maltodextrin fractions obtained from potato starch-derived maltodextrins (Paselli MD6) using ethanol precipitation methods.[4]

Biochem/physiol Actions

Pullulanase hydrolyzes α-1,6 linkages of starch.[1] Apart from pullulan, the enzyme also hydrolyzes α-1,6 linkages of amylopectin and glycogen. Maltose is the smallest sugar that it can release from an α-(1,6)-linkages.

Unit Definition

One unit will liberate 1.0 μmole of maltotriose (measured as glucose) from pullulan per min at pH 5.0 at 25 °C.

Physical form

Suspension in 3.2 M (NH4)2SO4 solution, pH 6.2

Preparation Note

Highly purified by a modification of ion exchange chromatography.

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fractionation of maltodextrins by ethanol
Defloor I, et al.
Journal of Chromatography A, 803 (1-2), 103-109 (1998)
M A Finkelman et al.
Applied and environmental microbiology, 43(2), 483-485 (1982-02-01)
A simple radiometric microassay for extracellular polysaccharide elaboration by yeast-like cells of Aureobasidium pullulans was developed, based upon a procedure originally described by Catley (FEBS Lett. 20:174-176, 1972). Incorporation of [C]glucose into pullulan was linear with respect to time and
Pullulan content of the ethanol precipitate from fermented agro-industrial wastes.
Israilides CJ, et al.
Applied Microbiology and Biotechnology, 49(5), 613-617 (1998)
L Noah et al.
The Journal of nutrition, 128(6), 977-985 (1998-06-18)
Resistant starch (RS) is thought to be present in large amounts in legume seeds; however, it has never been quantified in healthy humans. RS from cooked (atmospheric pressure) white beans was quantified in humans and pigs, and characterized to explain
Ghina Ali et al.
Carbohydrate polymers, 93(1), 109-115 (2013-03-08)
Pullulanase is an enzyme that hydrolyses the α-1,6 linkages of pullulan (Pull) to produce maltotriose units. We studied the capacity of pullulanase to cleave its modified substrate: carboxymethylpullulan (CMPull), synthesized with two different degrees of substitution (DS=0.16 and 0.8). Size

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service