Skip to Content
MilliporeSigma

L8638

Long chain alkylamine controlled pore glass

120-200 mesh, nominal diameter 500 Å

Synonym(s):

LCAA-CPG

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352200
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


particle size

120-200 mesh

pore size

500 Å nominal diameter

Application

Long chain alkylamine controlled pore glass is used in chromatography. Long chain alkylamine controlled pore glass has been used to develop a novel method for the immobilization of nucleotides and for the synthesis of DNA oligonucleotides and their 3′- or (3′,5′)-conjugates.

Other Notes

Ligand is mixture of primary and secondary alkylamines, approx. 1:1.


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library