H2510
1-O-Palmityl-rac-glycerol
~99%
Synonym(s):
rac-1-Hexadecyl-glycerol, rac-Glycerol-1-hexadecyl ether, DL-α-Hexadecylglycerol
About This Item
Recommended Products
assay
~99%
storage temp.
−20°C
SMILES string
CCCCCCCCCCCCCCCCOCC(O)CO
InChI
1S/C19H40O3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-22-18-19(21)17-20/h19-21H,2-18H2,1H3
InChI key
OOWQBDFWEXAXPB-UHFFFAOYSA-N
Gene Information
rat ... Cnr1(25248)
Looking for similar products? Visit Product Comparison Guide
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service