Skip to Content
MilliporeSigma

E3754

Ethylamine solution

70% in H2O

Synonym(s):

Aminoethane, Monoethylamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C2H5NH2
CAS Number:
Molecular Weight:
45.08
PubChem Substance ID:
UNSPSC Code:
12352100
Beilstein/REAXYS Number:
505933
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


concentration

70% in H2O

density

0.81 g/mL at 20 °C

storage temp.

2-8°C

SMILES string

CCN

InChI

1S/C2H7N/c1-2-3/h2-3H2,1H3

InChI key

QUSNBJAOOMFDIB-UHFFFAOYSA-N



Still not finding the right product?

Explore all of our products under Ethylamine solution


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

71.6 °F

flash_point_c

22 °C

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library