Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABF1083

Sigma-Aldrich

Anti-IFIT5 Antibody, clone 10A12.1

clone 10A12.1, from mouse

Synonym(s):

Interferon-induced protein with tetratricopeptide repeats 5, IFIT-5, Interferon-induced 58 kDa protein, P58, Retinoic acid- and interferon-inducible 58 kDa protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

10A12.1, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... IFIT5(24138)

General description

Interferon-induced protein with tetratricopeptide repeats 5 (UniProt Q13325; also known as IFIT-5, Interferon-induced 58 kDa protein, P58, Retinoic acid- and interferon-inducible 58 kDa protein) is encoded by the IFIT5 (also known as ISG58, RI58) gene (Gene ID 24138) in human. IFIT1, IFIT2, IFIT3, and IFIT5 constitute the family of interferon-stimulated genes (ISGs) that are induced in response to virus infection, stimulation by interferons (IFNs) or double-stranded RNA (dsRNA). The protiens encoded by IFIT1 and IFIT2 are called ISG56 (p56) and ISG54 (p54) in both human and murine species, while the murine IFIT3 (ISG49 or p49) is of a different size than its human counterpart (ISG60 or p60) and no known murine IFIT5 analog exists. IFIT family proteins restrict virus replication by various means. IFIT1 is known to disrupt complex formation between eIF3 and HCV RNA and inhibit HPV E1 helicase. IFIT3 is shown to enhance TBK1 and MAVS interation to potentiate antiviral response. IFIT5 is reported to facilitate the interaction between I B kinase (IKK) and TGF- -activated kinase 1 (TAK1) by binding both proteins, resulting in enhanced NF- B activation as a reuslt of increased IKK phosphorylation and activation. In addition, both IFIT1 and IFIT5 are shown to recognize viral RNA carrying 5 -triphosphate (PPP-RNA). Human IFIT5 contains eight tetratricopeptide repeat (TPR) helix-turn-helix motifs (a.a. 51-84, 94-127, 138-173, 181-214, 249-282, 338-371, 376-410, 435-468), with a.a. 254-260 within TPR repeat 5 mediating PPP-RNA interaction.

Specificity

Clone 10A12.1 epitope lies within region encompassing TPR repeats 6 and 8 present in both spliced isoforms of human IFIT5 reported by UniProt (Q13325).

Immunogen

GST-tagged recombinant human IFIT5 fragment encompassing TPR repeats 6 and 8.

Application

Research Category
Inflammation & Immunology
This mouse monoclonal Anti-IFIT5, clone 10A12.1 Antibody, Cat. No. MABF1083 is validated for use in Western Blotting for the detection of IFIT5.

Quality

Evaluated by Western Blotting in THP-1 cell lysate.

Western Blotting Analysis: A 1:5,000 dilution of this antibody detected IFIT5 in 10 µg of THP-1 cell lysate.

Target description

~55/50 kDa observed. 55.85/50.52 kDa (isoform 1/2) calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified.
Purified mouse IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service