Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABT40

Sigma-Aldrich

Anti-MFAP-5 (MAGP2) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Microfibrillar-associated protein 5, MFAP-5, MP25, Microfibril-associated glycoprotein 2, MAGP-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MFAP5(8076)

General description

Microfibril-associated protein 2 (MAGP-2), also known as microfibrillar associated protein 5 (MFAP-5), is a multifunctional secreted protein that acts as a promoter of tumor cell survival and angiogenesis. Through alphaVbeta3 integrin-mediated signaling, it is suggested that MAGP2 may provide a potential mechanistic link between MAGP2 and angiogenesis, as well as patient survival. Based on a high level of homology between MAGP (MFAP-2) and MAGP-2, both proteins are shown to bind to and activate Notch-1, as well as mediate the subsequent release of the Notch-1 extracellular domain.

Immunogen

KLH-conjugated linear peptide corresponding to human MFAP-5 (MAGP-2).

Application

Research Category
Cell Structure
Research Sub Category
Glycobiology

Adhesion (CAMs)
This Anti-MFAP-5 (MAGP2) Antibody is validated for use in WB for the detection of MFAP-5 (MAGP2).

Quality

Evaluated by Western Blot in human skeletal muscle tissue lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected MFAP-5 (MAGP-2) in skeletal muscle tissue lysate.

Target description

~19 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human skeletal muscle tissue lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service